Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161875431-161875702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA1
Entrez Gene ID
2554
Gene Name
gamma-aminobutyric acid type A receptor alpha1 subunit
Gene Aliases
ECA4, EIEE19, EJM, EJM5
UniGene
Hs.175934
Gene Chromosome Location
Chr.5: 161847191-161899959 on Build GRCh38
Transcripts
NM_000806.5 NM_001127643.1 NM_001127644.1 NM_001127645.1 NM_001127648.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4978533 c.553G>A p.G185R Chr.5: 161875636-161875636 on Build GRCh38
1435744 c.501G>A p.P167P Chr.5: 161875584-161875584 on Build GRCh38
6319584 c.497G>T p.C166F Chr.5: 161875580-161875580 on Build GRCh38
5119570 c.549A>C p.K183N Chr.5: 161875632-161875632 on Build GRCh38
1065320 c.529G>A p.A177T Chr.5: 161875612-161875612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747735903 Chr.5: 161875629-161875629 on Build GRCh38
rs555755562 Chr.5: 161875525-161875525 on Build GRCh38
rs35223687 Chr.5: 161875601-161875602 on Build GRCh38
rs746503521 Chr.5: 161875509-161875509 on Build GRCh38
rs945438671 Chr.5: 161875620-161875620 on Build GRCh38
rs138145692 Chr.5: 161875604-161875605 on Build GRCh38
rs762857527 Chr.5: 161875529-161875529 on Build GRCh38
rs956557281 Chr.5: 161875507-161875507 on Build GRCh38
rs762141453 Chr.5: 161875458-161875458 on Build GRCh38
rs769309543 Chr.5: 161875647-161875647 on Build GRCh38
rs764106229 Chr.5: 161875535-161875535 on Build GRCh38
rs911229801 Chr.5: 161875541-161875541 on Build GRCh38
rs909004879 Chr.5: 161875504-161875504 on Build GRCh38
rs753939740 Chr.5: 161875543-161875543 on Build GRCh38
rs34281832 Chr.5: 161875492-161875493 on Build GRCh38
rs984824004 Chr.5: 161875501-161875501 on Build GRCh38
rs770649706 Chr.5: 161875655-161875655 on Build GRCh38
rs772977557 Chr.5: 161875652-161875652 on Build GRCh38
rs768345960 Chr.5: 161875514-161875514 on Build GRCh38
rs1030813639 Chr.5: 161875503-161875503 on Build GRCh38
rs41311753 Chr.5: 161875530-161875530 on Build GRCh38
rs780700439 Chr.5: 161875612-161875612 on Build GRCh38
rs762653176 Chr.5: 161875653-161875653 on Build GRCh38
rs573982851 Chr.5: 161875532-161875532 on Build GRCh38
rs765361211 Chr.5: 161875671-161875671 on Build GRCh38
rs796052499 Chr.5: 161875567-161875567 on Build GRCh38
rs767478000 Chr.5: 161875660-161875660 on Build GRCh38
rs761997367 Chr.5: 161875670-161875670 on Build GRCh38
rs769299843 Chr.5: 161875623-161875623 on Build GRCh38
rs757378039 Chr.5: 161875546-161875546 on Build GRCh38
rs771228034 Chr.5: 161875545-161875546 on Build GRCh38
rs780800247 Chr.5: 161875626-161875626 on Build GRCh38
rs183274236 Chr.5: 161875524-161875524 on Build GRCh38
rs774147411 Chr.5: 161875656-161875656 on Build GRCh38
rs779231183 Chr.5: 161875555-161875555 on Build GRCh38
rs796052489 Chr.5: 161875589-161875589 on Build GRCh38
rs777148533 Chr.5: 161875663-161875663 on Build GRCh38
rs200750234 Chr.5: 161875584-161875584 on Build GRCh38
rs367546876 Chr.5: 161875485-161875485 on Build GRCh38
rs772852609 Chr.5: 161875526-161875526 on Build GRCh38
rs750728250 Chr.5: 161875583-161875583 on Build GRCh38
rs759425091 Chr.5: 161875658-161875658 on Build GRCh38
rs776285228 Chr.5: 161875520-161875520 on Build GRCh38
rs747452052 Chr.5: 161875614-161875614 on Build GRCh38
rs753826414 Chr.5: 161875665-161875665 on Build GRCh38

View Full Product Details