Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 133356357-133356610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC4
Entrez Gene ID
60412
Gene Name
exocyst complex component 4
Gene Aliases
SEC8, SEC8L1, Sec8p
UniGene
Hs.321273
Gene Chromosome Location
Chr.7: 133253067-134067137 on Build GRCh38
Transcripts
NM_001037126.1 NM_021807.3 XM_005250523.4 XM_017012495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3633527 c.876C>T p.I292I Chr.7: 133356442-133356442 on Build GRCh38
6259141 c.955G>T p.A319S Chr.7: 133356521-133356521 on Build GRCh38
3730349 c.791A>C p.K264T Chr.7: 133356357-133356357 on Build GRCh38
4359880 c.985G>A p.V329I Chr.7: 133356551-133356551 on Build GRCh38
4956992 c.847A>C p.I283L Chr.7: 133356413-133356413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527817629 Chr.7: 133356448-133356448 on Build GRCh38
rs768071863 Chr.7: 133356496-133356496 on Build GRCh38
rs754080176 Chr.7: 133356420-133356420 on Build GRCh38
rs374961055 Chr.7: 133356513-133356513 on Build GRCh38
rs766479473 Chr.7: 133356512-133356512 on Build GRCh38
rs761147075 Chr.7: 133356508-133356508 on Build GRCh38
rs202172178 Chr.7: 133356550-133356550 on Build GRCh38
rs761216417 Chr.7: 133356565-133356565 on Build GRCh38
rs200156091 Chr.7: 133356522-133356522 on Build GRCh38
rs762386688 Chr.7: 133356385-133356385 on Build GRCh38
rs201609585 Chr.7: 133356539-133356539 on Build GRCh38
rs750491430 Chr.7: 133356404-133356404 on Build GRCh38
rs765520328 Chr.7: 133356500-133356501 on Build GRCh38
rs751285429 Chr.7: 133356528-133356528 on Build GRCh38
rs143291638 Chr.7: 133356409-133356409 on Build GRCh38
rs944286544 Chr.7: 133356432-133356432 on Build GRCh38
rs750982913 Chr.7: 133356505-133356505 on Build GRCh38
rs369312406 Chr.7: 133356540-133356540 on Build GRCh38
rs768161600 Chr.7: 133356390-133356390 on Build GRCh38
rs768384421 Chr.7: 133356542-133356542 on Build GRCh38
rs141180561 Chr.7: 133356541-133356541 on Build GRCh38
rs141369405 Chr.7: 133356509-133356509 on Build GRCh38
rs770811321 Chr.7: 133356427-133356427 on Build GRCh38
rs753908169 Chr.7: 133356521-133356521 on Build GRCh38
rs772554686 Chr.7: 133356551-133356551 on Build GRCh38
rs755224502 Chr.7: 133356421-133356421 on Build GRCh38
rs774808256 Chr.7: 133356470-133356470 on Build GRCh38
rs77094376 Chr.7: 133356546-133356546 on Build GRCh38
rs777516999 Chr.7: 133356424-133356424 on Build GRCh38
rs139031553 Chr.7: 133356419-133356419 on Build GRCh38
rs545267730 Chr.7: 133356426-133356426 on Build GRCh38
rs140636237 Chr.7: 133356465-133356465 on Build GRCh38
rs868537272 Chr.7: 133356492-133356492 on Build GRCh38
rs762300660 Chr.7: 133356471-133356471 on Build GRCh38
rs139150672 Chr.7: 133356571-133356571 on Build GRCh38
rs35224977 Chr.7: 133356442-133356442 on Build GRCh38
rs769331521 Chr.7: 133356457-133356457 on Build GRCh38
rs762634261 Chr.7: 133356462-133356462 on Build GRCh38
rs766848115 Chr.7: 133356569-133356569 on Build GRCh38
rs931505556 Chr.7: 133356449-133356449 on Build GRCh38
rs370517844 Chr.7: 133356562-133356562 on Build GRCh38
rs147214615 Chr.7: 133356461-133356461 on Build GRCh38

View Full Product Details