Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76008647-76008872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
UniGene
Hs.500635
Gene Chromosome Location
Chr.17: 76006845-76027452 on Build GRCh38
Transcripts
NM_001320747.1 NM_001988.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5530656 c.4534G>A p.A1512T Chr.17: 76008671-76008671 on Build GRCh38
1284788 c.4395G>C p.V1465V Chr.17: 76008810-76008810 on Build GRCh38
6322205 c.4554C>A p.T1518T Chr.17: 76008651-76008651 on Build GRCh38
4069759 c.4479C>T p.T1493T Chr.17: 76008726-76008726 on Build GRCh38
4069760 c.4356G>A p.R1452R Chr.17: 76008849-76008849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766819423 Chr.17: 76008755-76008755 on Build GRCh38
rs373960086 Chr.17: 76008716-76008716 on Build GRCh38
rs778262916 Chr.17: 76008798-76008798 on Build GRCh38
rs756740512 Chr.17: 76008792-76008792 on Build GRCh38
rs755513899 Chr.17: 76008760-76008760 on Build GRCh38
rs748770869 Chr.17: 76008722-76008722 on Build GRCh38
rs755349309 Chr.17: 76008695-76008695 on Build GRCh38
rs770036148 Chr.17: 76008830-76008830 on Build GRCh38
rs766845248 Chr.17: 76008686-76008686 on Build GRCh38
rs528132734 Chr.17: 76008688-76008688 on Build GRCh38
rs150027671 Chr.17: 76008683-76008683 on Build GRCh38
rs550275335 Chr.17: 76008831-76008831 on Build GRCh38
rs775277575 Chr.17: 76008821-76008821 on Build GRCh38
rs200073444 Chr.17: 76008671-76008671 on Build GRCh38
rs148808819 Chr.17: 76008726-76008726 on Build GRCh38
rs1055303658 Chr.17: 76008771-76008771 on Build GRCh38
rs765385227 Chr.17: 76008742-76008742 on Build GRCh38
rs770775128 Chr.17: 76008723-76008723 on Build GRCh38
rs144390976 Chr.17: 76008684-76008684 on Build GRCh38
rs781674876 Chr.17: 76008701-76008701 on Build GRCh38
rs775233961 Chr.17: 76008738-76008738 on Build GRCh38
rs763066973 Chr.17: 76008840-76008840 on Build GRCh38
rs773171068 Chr.17: 76008748-76008748 on Build GRCh38
rs199551812 Chr.17: 76008777-76008777 on Build GRCh38
rs751902650 Chr.17: 76008689-76008689 on Build GRCh38
rs753094081 Chr.17: 76008772-76008772 on Build GRCh38
rs746741512 Chr.17: 76008827-76008827 on Build GRCh38
rs745858072 Chr.17: 76008812-76008812 on Build GRCh38
rs774293924 Chr.17: 76008851-76008851 on Build GRCh38
rs962803888 Chr.17: 76008713-76008713 on Build GRCh38
rs138016423 Chr.17: 76008758-76008758 on Build GRCh38
rs930188197 Chr.17: 76008751-76008751 on Build GRCh38
rs1046226109 Chr.17: 76008829-76008829 on Build GRCh38
rs113135904 Chr.17: 76008847-76008847 on Build GRCh38
rs768065780 Chr.17: 76008761-76008761 on Build GRCh38
rs771992734 Chr.17: 76008820-76008820 on Build GRCh38
rs763049808 Chr.17: 76008752-76008752 on Build GRCh38
rs199961325 Chr.17: 76008789-76008789 on Build GRCh38
rs747847879 Chr.17: 76008714-76008714 on Build GRCh38
rs144924893 Chr.17: 76008736-76008736 on Build GRCh38
rs762089155 Chr.17: 76008740-76008740 on Build GRCh38
rs777479943 Chr.17: 76008720-76008720 on Build GRCh38
rs894868698 Chr.17: 76008781-76008781 on Build GRCh38
rs918846343 Chr.17: 76008745-76008745 on Build GRCh38
rs372237819 Chr.17: 76008739-76008739 on Build GRCh38

View Full Product Details