Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10701739-10701994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YBX3
Entrez Gene ID
8531
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA, CSDA1, DBPA, ZONAB
UniGene
Hs.221889
Gene Chromosome Location
Chr.12: 10699077-10723448 on Build GRCh38
Transcripts
NM_001145426.1 NM_003651.4 XM_017020122.1 XM_017020123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
136050 c.1033C>T p.P345S Chr.12: 10701980-10701980 on Build GRCh38
467732 c.1050A>T p.K350N Chr.12: 10701963-10701963 on Build GRCh38
7126954 c.1027A>G p.N343D Chr.12: 10701986-10701986 on Build GRCh38
1298874 c.1019G>A p.R340H Chr.12: 10701994-10701994 on Build GRCh38
6050546 c.1030G>A p.A344T Chr.12: 10701983-10701983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150858609 Chr.12: 10701772-10701772 on Build GRCh38
rs776343916 Chr.12: 10701926-10701926 on Build GRCh38
rs769094490 Chr.12: 10701913-10701913 on Build GRCh38
rs763177307 Chr.12: 10701912-10701912 on Build GRCh38
rs966952528 Chr.12: 10701964-10701964 on Build GRCh38
rs949521652 Chr.12: 10701856-10701856 on Build GRCh38
rs959573903 Chr.12: 10701821-10701821 on Build GRCh38
rs773997903 Chr.12: 10701925-10701925 on Build GRCh38
rs2305850 Chr.12: 10701852-10701852 on Build GRCh38
rs868203261 Chr.12: 10701788-10701788 on Build GRCh38
rs372517831 Chr.12: 10701918-10701918 on Build GRCh38
rs759190470 Chr.12: 10701931-10701931 on Build GRCh38
rs762456118 Chr.12: 10701919-10701919 on Build GRCh38
rs559202899 Chr.12: 10701930-10701930 on Build GRCh38
rs765970100 Chr.12: 10701924-10701924 on Build GRCh38
rs991405570 Chr.12: 10701826-10701826 on Build GRCh38
rs887003608 Chr.12: 10701792-10701792 on Build GRCh38
rs541600275 Chr.12: 10701969-10701969 on Build GRCh38
rs373361738 Chr.12: 10701939-10701939 on Build GRCh38
rs75048880 Chr.12: 10701806-10701806 on Build GRCh38
rs111863508 Chr.12: 10701893-10701893 on Build GRCh38
rs376817541 Chr.12: 10701936-10701936 on Build GRCh38
rs781245935 Chr.12: 10701955-10701955 on Build GRCh38
rs751477844 Chr.12: 10701940-10701940 on Build GRCh38
rs745877932 Chr.12: 10701958-10701958 on Build GRCh38
rs1021176328 Chr.12: 10701949-10701949 on Build GRCh38
rs949685631 Chr.12: 10701779-10701779 on Build GRCh38
rs939864330 Chr.12: 10701802-10701802 on Build GRCh38
rs759094510 Chr.12: 10701927-10701927 on Build GRCh38
rs764743148 Chr.12: 10701928-10701928 on Build GRCh38
rs376512201 Chr.12: 10701941-10701941 on Build GRCh38
rs2305849 Chr.12: 10701938-10701938 on Build GRCh38
rs898578596 Chr.12: 10701851-10701851 on Build GRCh38
rs980010460 Chr.12: 10701970-10701970 on Build GRCh38
rs931330932 Chr.12: 10701965-10701965 on Build GRCh38
rs576756214 Chr.12: 10701820-10701820 on Build GRCh38

View Full Product Details