Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93870506-93870755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP2
Entrez Gene ID
30836
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2
UniGene
Hs.601998
Gene Chromosome Location
Chr.1: 93869458-93879206 on Build GRCh38
Transcripts
NM_014597.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806142 c.2135G>T p.R712I Chr.1: 93870725-93870725 on Build GRCh38
6460796 c.2174G>A p.R725K Chr.1: 93870686-93870686 on Build GRCh38
7296153 c.2159C>T p.A720V Chr.1: 93870701-93870701 on Build GRCh38
5570304 c.2139T>C p.T713T Chr.1: 93870721-93870721 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs149042261 Chr.1: 93870708-93870708 on Build GRCh38
rs1056903261 Chr.1: 93870581-93870581 on Build GRCh38
rs761505126 Chr.1: 93870664-93870664 on Build GRCh38
rs899635460 Chr.1: 93870549-93870549 on Build GRCh38
rs777846665 Chr.1: 93870729-93870729 on Build GRCh38
rs891933474 Chr.1: 93870677-93870677 on Build GRCh38
rs1011844327 Chr.1: 93870559-93870559 on Build GRCh38
rs761881995 Chr.1: 93870587-93870590 on Build GRCh38
rs762729170 Chr.1: 93870643-93870643 on Build GRCh38
rs968133667 Chr.1: 93870603-93870603 on Build GRCh38
rs1022340886 Chr.1: 93870595-93870595 on Build GRCh38
rs774257877 Chr.1: 93870658-93870658 on Build GRCh38
rs755996360 Chr.1: 93870705-93870705 on Build GRCh38
rs752705165 Chr.1: 93870720-93870720 on Build GRCh38
rs747474951 Chr.1: 93870730-93870730 on Build GRCh38
rs1008122721 Chr.1: 93870534-93870534 on Build GRCh38
rs376226397 Chr.1: 93870651-93870651 on Build GRCh38
rs146863861 Chr.1: 93870588-93870588 on Build GRCh38
rs532855425 Chr.1: 93870623-93870623 on Build GRCh38
rs758634381 Chr.1: 93870725-93870725 on Build GRCh38
rs750306219 Chr.1: 93870695-93870695 on Build GRCh38
rs767526290 Chr.1: 93870673-93870673 on Build GRCh38
rs1018510753 Chr.1: 93870541-93870541 on Build GRCh38

View Full Product Details