Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133527341-133527614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2E1
Entrez Gene ID
1571
Gene Name
cytochrome P450 family 2 subfamily E member 1
Gene Aliases
CPE1, CYP2E, P450-J, P450C2E
UniGene
Hs.12907
Gene Chromosome Location
Chr.10: 133527363-133539116 on Build GRCh38
Transcripts
NM_000773.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268976 c.41C>T p.A14V Chr.10: 133527436-133527436 on Build GRCh38
1703288 c.112C>T p.P38S Chr.10: 133527507-133527507 on Build GRCh38
1703287 c.62C>T p.S21F Chr.10: 133527457-133527457 on Build GRCh38
4675953 c.42G>A p.A14A Chr.10: 133527437-133527437 on Build GRCh38
356437 c.19A>T p.T7S Chr.10: 133527414-133527414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397709571 Chr.10: 133527363-133527364 on Build GRCh38
rs756596289 Chr.10: 133527564-133527564 on Build GRCh38
rs72559709 Chr.10: 133527554-133527555 on Build GRCh38
rs764387026 Chr.10: 133527419-133527419 on Build GRCh38
rs766977009 Chr.10: 133527387-133527387 on Build GRCh38
rs76379844 Chr.10: 133527573-133527573 on Build GRCh38
rs771631693 Chr.10: 133527577-133527577 on Build GRCh38
rs527949682 Chr.10: 133527527-133527527 on Build GRCh38
rs900694954 Chr.10: 133527459-133527459 on Build GRCh38
rs200292747 Chr.10: 133527364-133527364 on Build GRCh38
rs543066971 Chr.10: 133527426-133527426 on Build GRCh38
rs369847743 Chr.10: 133527537-133527537 on Build GRCh38
rs367957731 Chr.10: 133527444-133527444 on Build GRCh38
rs766143856 Chr.10: 133527395-133527395 on Build GRCh38
rs979091921 Chr.10: 133527514-133527514 on Build GRCh38
rs769059259 Chr.10: 133527472-133527472 on Build GRCh38
rs761293223 Chr.10: 133527383-133527383 on Build GRCh38
rs776753984 Chr.10: 133527518-133527518 on Build GRCh38
rs750499891 Chr.10: 133527556-133527556 on Build GRCh38
rs780517566 Chr.10: 133527567-133527567 on Build GRCh38
rs747781058 Chr.10: 133527575-133527575 on Build GRCh38
rs771599544 Chr.10: 133527504-133527504 on Build GRCh38
rs757352167 Chr.10: 133527433-133527433 on Build GRCh38
rs201068922 Chr.10: 133527452-133527452 on Build GRCh38
rs925010450 Chr.10: 133527533-133527533 on Build GRCh38
rs371502619 Chr.10: 133527368-133527368 on Build GRCh38
rs1037280829 Chr.10: 133527579-133527579 on Build GRCh38
rs869155073 Chr.10: 133527364-133527365 on Build GRCh38
rs753713804 Chr.10: 133527405-133527405 on Build GRCh38
rs114535864 Chr.10: 133527569-133527569 on Build GRCh38
rs780358119 Chr.10: 133527451-133527451 on Build GRCh38
rs375169910 Chr.10: 133527437-133527437 on Build GRCh38
rs757431552 Chr.10: 133527543-133527543 on Build GRCh38
rs143746211 Chr.10: 133527550-133527550 on Build GRCh38
rs563043306 Chr.10: 133527436-133527436 on Build GRCh38
rs775845751 Chr.10: 133527366-133527366 on Build GRCh38
rs772430702 Chr.10: 133527461-133527461 on Build GRCh38
rs775675704 Chr.10: 133527477-133527477 on Build GRCh38
rs759372423 Chr.10: 133527522-133527522 on Build GRCh38
rs201519230 Chr.10: 133527365-133527365 on Build GRCh38
rs773200181 Chr.10: 133527390-133527390 on Build GRCh38
rs374647442 Chr.10: 133527367-133527367 on Build GRCh38
rs958141259 Chr.10: 133527562-133527562 on Build GRCh38
rs779311330 Chr.10: 133527571-133527571 on Build GRCh38
rs137875795 Chr.10: 133527509-133527509 on Build GRCh38
rs72559707 Chr.10: 133527505-133527506 on Build GRCh38
rs773963522 Chr.10: 133527478-133527478 on Build GRCh38
rs766438198 Chr.10: 133527515-133527515 on Build GRCh38
rs747825060 Chr.10: 133527501-133527501 on Build GRCh38
rs997759133 Chr.10: 133527463-133527463 on Build GRCh38
rs758640808 Chr.10: 133527407-133527407 on Build GRCh38
rs59868347 Chr.10: 133527471-133527471 on Build GRCh38
rs11445593 Chr.10: 133527362-133527363 on Build GRCh38
rs76271067 Chr.10: 133527508-133527508 on Build GRCh38
rs577328423 Chr.10: 133527371-133527371 on Build GRCh38

View Full Product Details