Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74719148-74719409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZ
Entrez Gene ID
1429
Gene Name
crystallin zeta
Gene Aliases
-
UniGene
Hs.83114
Gene Chromosome Location
Chr.1: 74705482-74733408 on Build GRCh38
Transcripts
NM_001130042.1 NM_001130043.1 NM_001134759.1 NM_001889.3 XM_005270491.4 XM_011540747.1 XM_017000367.1 XM_017000368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6380363 c.293C>A p.T98K Chr.1: 74719344-74719344 on Build GRCh38
160225 c.271G>A p.D91N Chr.1: 74719366-74719366 on Build GRCh38
7229842 c.303G>A p.G101G Chr.1: 74719334-74719334 on Build GRCh38
3419458 c.412C>T p.R138* Chr.1: 74719225-74719225 on Build GRCh38
7078543 c.413G>T p.R138L Chr.1: 74719224-74719224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763455323 Chr.1: 74719284-74719284 on Build GRCh38
rs780868788 Chr.1: 74719252-74719252 on Build GRCh38
rs751875474 Chr.1: 74719303-74719303 on Build GRCh38
rs1037440251 Chr.1: 74719249-74719249 on Build GRCh38
rs142737449 Chr.1: 74719297-74719297 on Build GRCh38
rs763394914 Chr.1: 74719177-74719177 on Build GRCh38
rs202197566 Chr.1: 74719306-74719306 on Build GRCh38
rs997035065 Chr.1: 74719205-74719205 on Build GRCh38
rs750607276 Chr.1: 74719312-74719312 on Build GRCh38
rs200151638 Chr.1: 74719186-74719186 on Build GRCh38
rs780872649 Chr.1: 74719353-74719353 on Build GRCh38
rs750648473 Chr.1: 74719211-74719211 on Build GRCh38
rs973305542 Chr.1: 74719377-74719377 on Build GRCh38
rs149889365 Chr.1: 74719365-74719365 on Build GRCh38
rs757098166 Chr.1: 74719349-74719349 on Build GRCh38
rs139024113 Chr.1: 74719334-74719334 on Build GRCh38
rs754914441 Chr.1: 74719286-74719286 on Build GRCh38
rs769338292 Chr.1: 74719368-74719368 on Build GRCh38
rs752638968 Chr.1: 74719223-74719223 on Build GRCh38
rs751929973 Chr.1: 74719193-74719193 on Build GRCh38
rs201156382 Chr.1: 74719195-74719195 on Build GRCh38
rs575610327 Chr.1: 74719219-74719219 on Build GRCh38
rs746787769 Chr.1: 74719247-74719247 on Build GRCh38
rs777849852 Chr.1: 74719344-74719344 on Build GRCh38
rs764404931 Chr.1: 74719180-74719180 on Build GRCh38
rs769167696 Chr.1: 74719291-74719291 on Build GRCh38
rs755089362 Chr.1: 74719376-74719376 on Build GRCh38
rs371028828 Chr.1: 74719347-74719347 on Build GRCh38
rs760836369 Chr.1: 74719327-74719327 on Build GRCh38
rs542527429 Chr.1: 74719250-74719250 on Build GRCh38
rs57504503 Chr.1: 74719208-74719208 on Build GRCh38
rs540399244 Chr.1: 74719343-74719343 on Build GRCh38
rs766355707 Chr.1: 74719179-74719179 on Build GRCh38
rs776044518 Chr.1: 74719175-74719175 on Build GRCh38
rs758414819 Chr.1: 74719224-74719224 on Build GRCh38
rs904340409 Chr.1: 74719348-74719348 on Build GRCh38
rs768190068 Chr.1: 74719376-74719376 on Build GRCh38
rs764696127 Chr.1: 74719174-74719174 on Build GRCh38
rs139734636 Chr.1: 74719373-74719373 on Build GRCh38
rs777653390 Chr.1: 74719225-74719225 on Build GRCh38
rs774897599 Chr.1: 74719283-74719283 on Build GRCh38
rs767717411 Chr.1: 74719209-74719209 on Build GRCh38
rs147181596 Chr.1: 74719270-74719270 on Build GRCh38
rs766414010 Chr.1: 74719332-74719332 on Build GRCh38
rs779563277 Chr.1: 74719369-74719369 on Build GRCh38
rs756193644 Chr.1: 74719217-74719217 on Build GRCh38
rs554764859 Chr.1: 74719262-74719262 on Build GRCh38

View Full Product Details