Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29619544-29619811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO6
Entrez Gene ID
84940
Gene Name
coronin 6
Gene Aliases
-
UniGene
Hs.143046
Gene Chromosome Location
Chr.17: 29614756-29622912 on Build GRCh38
Transcripts
NM_032854.3 XM_005258047.3 XM_005258048.4 XM_011525385.2 XM_011525386.2 XM_011525387.2 XM_011525388.2 XM_011525389.2 XM_017025235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1248989 c.306C>T p.D102D Chr.17: 29619666-29619666 on Build GRCh38
5556048 c.236G>T p.G79V Chr.17: 29619736-29619736 on Build GRCh38
2802005 c.205C>T p.R69* Chr.17: 29619767-29619767 on Build GRCh38
6404704 c.218A>G p.N73S Chr.17: 29619754-29619754 on Build GRCh38
6426078 c.211G>T p.D71Y Chr.17: 29619761-29619761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760613230 Chr.17: 29619735-29619735 on Build GRCh38
rs765504035 Chr.17: 29619767-29619767 on Build GRCh38
rs758461627 Chr.17: 29619687-29619687 on Build GRCh38
rs538399226 Chr.17: 29619740-29619740 on Build GRCh38
rs771226997 Chr.17: 29619697-29619697 on Build GRCh38
rs893512419 Chr.17: 29619582-29619582 on Build GRCh38
rs780256611 Chr.17: 29619786-29619788 on Build GRCh38
rs746196281 Chr.17: 29619705-29619705 on Build GRCh38
rs762622246 Chr.17: 29619609-29619609 on Build GRCh38
rs372494637 Chr.17: 29619681-29619681 on Build GRCh38
rs781567271 Chr.17: 29619702-29619702 on Build GRCh38
rs763535509 Chr.17: 29619626-29619626 on Build GRCh38
rs774053469 Chr.17: 29619637-29619637 on Build GRCh38
rs754029706 Chr.17: 29619666-29619666 on Build GRCh38
rs755280721 Chr.17: 29619678-29619678 on Build GRCh38
rs972132363 Chr.17: 29619752-29619752 on Build GRCh38
rs528781661 Chr.17: 29619583-29619583 on Build GRCh38
rs753712051 Chr.17: 29619745-29619745 on Build GRCh38
rs761465111 Chr.17: 29619719-29619719 on Build GRCh38
rs193135655 Chr.17: 29619778-29619778 on Build GRCh38
rs963398554 Chr.17: 29619746-29619746 on Build GRCh38
rs774141399 Chr.17: 29619715-29619715 on Build GRCh38
rs11650514 Chr.17: 29619668-29619668 on Build GRCh38
rs1013203657 Chr.17: 29619686-29619686 on Build GRCh38
rs766158357 Chr.17: 29619661-29619661 on Build GRCh38
rs988372964 Chr.17: 29619589-29619589 on Build GRCh38
rs546908504 Chr.17: 29619584-29619584 on Build GRCh38
rs747479235 Chr.17: 29619696-29619696 on Build GRCh38
rs753662812 Chr.17: 29619762-29619762 on Build GRCh38
rs569893234 Chr.17: 29619604-29619604 on Build GRCh38
rs761374794 Chr.17: 29619651-29619651 on Build GRCh38
rs571811449 Chr.17: 29619653-29619653 on Build GRCh38
rs201624665 Chr.17: 29619656-29619656 on Build GRCh38
rs375419798 Chr.17: 29619680-29619680 on Build GRCh38
rs903333300 Chr.17: 29619612-29619612 on Build GRCh38
rs773314445 Chr.17: 29619729-29619729 on Build GRCh38
rs768393522 Chr.17: 29619710-29619710 on Build GRCh38
rs914166875 Chr.17: 29619652-29619652 on Build GRCh38
rs771642925 Chr.17: 29619727-29619727 on Build GRCh38
rs778264119 Chr.17: 29619693-29619693 on Build GRCh38
rs752856330 Chr.17: 29619769-29619769 on Build GRCh38

View Full Product Details