Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65389366-65389593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD8
Entrez Gene ID
83786
Gene Name
FERM domain containing 8
Gene Aliases
FKSG44
UniGene
Hs.578433
Gene Chromosome Location
Chr.11: 65386570-65413525 on Build GRCh38
Transcripts
NM_001300832.1 NM_001300833.1 NM_031904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685662 c.178G>A p.A60T Chr.11: 65389453-65389453 on Build GRCh38
7297657 c.101T>C p.V34A Chr.11: 65389376-65389376 on Build GRCh38
293457 c.186C>T p.R62R Chr.11: 65389461-65389461 on Build GRCh38
7130967 c.115G>A p.D39N Chr.11: 65389390-65389390 on Build GRCh38
5980113 c.184C>T p.R62C Chr.11: 65389459-65389459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757071534 Chr.11: 65389423-65389424 on Build GRCh38
rs767941954 Chr.11: 65389505-65389505 on Build GRCh38
rs758310158 Chr.11: 65389451-65389451 on Build GRCh38
rs532210089 Chr.11: 65389443-65389443 on Build GRCh38
rs757166148 Chr.11: 65389572-65389572 on Build GRCh38
rs763305900 Chr.11: 65389479-65389479 on Build GRCh38
rs975982203 Chr.11: 65389481-65389481 on Build GRCh38
rs768757210 Chr.11: 65389450-65389450 on Build GRCh38
rs752130559 Chr.11: 65389431-65389431 on Build GRCh38
rs764550211 Chr.11: 65389427-65389427 on Build GRCh38
rs200659044 Chr.11: 65389573-65389573 on Build GRCh38
rs199675557 Chr.11: 65389403-65389403 on Build GRCh38
rs200245458 Chr.11: 65389394-65389394 on Build GRCh38
rs548185455 Chr.11: 65389527-65389527 on Build GRCh38
rs528421662 Chr.11: 65389506-65389506 on Build GRCh38
rs750477529 Chr.11: 65389440-65389440 on Build GRCh38
rs775042781 Chr.11: 65389478-65389478 on Build GRCh38
rs923883704 Chr.11: 65389485-65389485 on Build GRCh38
rs751430286 Chr.11: 65389554-65389554 on Build GRCh38
rs774715033 Chr.11: 65389500-65389500 on Build GRCh38
rs11552893 Chr.11: 65389426-65389426 on Build GRCh38
rs563105598 Chr.11: 65389437-65389437 on Build GRCh38
rs146504901 Chr.11: 65389461-65389461 on Build GRCh38
rs746885699 Chr.11: 65389453-65389453 on Build GRCh38
rs757317982 Chr.11: 65389436-65389436 on Build GRCh38
rs140843850 Chr.11: 65389493-65389493 on Build GRCh38
rs947804202 Chr.11: 65389561-65389561 on Build GRCh38
rs190868663 Chr.11: 65389400-65389400 on Build GRCh38
rs76814425 Chr.11: 65389395-65389395 on Build GRCh38
rs745388067 Chr.11: 65389460-65389460 on Build GRCh38
rs576827620 Chr.11: 65389456-65389456 on Build GRCh38
rs760718536 Chr.11: 65389517-65389517 on Build GRCh38
rs763354731 Chr.11: 65389421-65389421 on Build GRCh38
rs373521065 Chr.11: 65389459-65389459 on Build GRCh38
rs371990682 Chr.11: 65389503-65389503 on Build GRCh38
rs992420123 Chr.11: 65389539-65389539 on Build GRCh38
rs144150822 Chr.11: 65389452-65389452 on Build GRCh38
rs375533419 Chr.11: 65389386-65389386 on Build GRCh38
rs780162209 Chr.11: 65389445-65389445 on Build GRCh38
rs766594002 Chr.11: 65389520-65389520 on Build GRCh38
rs769069315 Chr.11: 65389486-65389486 on Build GRCh38
rs1014585740 Chr.11: 65389458-65389458 on Build GRCh38
rs775870773 Chr.11: 65389404-65389404 on Build GRCh38
rs183893280 Chr.11: 65389546-65389546 on Build GRCh38
rs931788549 Chr.11: 65389502-65389502 on Build GRCh38
rs767579526 Chr.11: 65389439-65389439 on Build GRCh38
rs755263485 Chr.11: 65389550-65389550 on Build GRCh38
rs201594600 Chr.11: 65389495-65389495 on Build GRCh38
rs751379042 Chr.11: 65389406-65389406 on Build GRCh38
rs777645233 Chr.11: 65389553-65389553 on Build GRCh38

View Full Product Details