Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3858613-3858872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFB
Entrez Gene ID
1677
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
CAD, CPAN, DFF-40, DFF2, DFF40
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3857143-3885429 on Build GRCh38
Transcripts
NM_001282669.1 NM_001320132.1 NM_001320136.1 NM_004402.3 NR_104222.1 NR_135150.1 NR_135151.1 NR_135152.1 XM_011540865.2 XM_017000498.1 XM_017000499.1 XM_017000500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900822 c.208G>T p.V70L Chr.1: 3858811-3858811 on Build GRCh38
4885593 c.202G>A p.E68K Chr.1: 3858805-3858805 on Build GRCh38
6642770 c.159G>A p.T53T Chr.1: 3858762-3858762 on Build GRCh38
4940004 c.209T>G p.V70G Chr.1: 3858812-3858812 on Build GRCh38
908977 c.126C>T p.R42R Chr.1: 3858729-3858729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772537088 Chr.1: 3858771-3858771 on Build GRCh38
rs750659695 Chr.1: 3858772-3858772 on Build GRCh38
rs932632147 Chr.1: 3858815-3858815 on Build GRCh38
rs147517965 Chr.1: 3858817-3858817 on Build GRCh38
rs373269179 Chr.1: 3858707-3858707 on Build GRCh38
rs370380322 Chr.1: 3858672-3858672 on Build GRCh38
rs983682958 Chr.1: 3858715-3858715 on Build GRCh38
rs749809863 Chr.1: 3858697-3858697 on Build GRCh38
rs779628146 Chr.1: 3858780-3858780 on Build GRCh38
rs1022608172 Chr.1: 3858638-3858638 on Build GRCh38
rs779542927 Chr.1: 3858678-3858679 on Build GRCh38
rs757395687 Chr.1: 3858763-3858763 on Build GRCh38
rs755440154 Chr.1: 3858735-3858735 on Build GRCh38
rs143919670 Chr.1: 3858822-3858822 on Build GRCh38
rs766199794 Chr.1: 3858749-3858749 on Build GRCh38
rs769029152 Chr.1: 3858777-3858777 on Build GRCh38
rs749660067 Chr.1: 3858775-3858775 on Build GRCh38
rs772779769 Chr.1: 3858793-3858793 on Build GRCh38
rs775465576 Chr.1: 3858847-3858847 on Build GRCh38
rs139312649 Chr.1: 3858784-3858784 on Build GRCh38
rs773056030 Chr.1: 3858734-3858734 on Build GRCh38
rs927393456 Chr.1: 3858792-3858792 on Build GRCh38
rs58941669 Chr.1: 3858662-3858662 on Build GRCh38
rs1018676256 Chr.1: 3858658-3858658 on Build GRCh38
rs780229377 Chr.1: 3858690-3858690 on Build GRCh38
rs150699255 Chr.1: 3858773-3858773 on Build GRCh38
rs367946340 Chr.1: 3858836-3858836 on Build GRCh38
rs767846243 Chr.1: 3858795-3858795 on Build GRCh38
rs920264907 Chr.1: 3858705-3858705 on Build GRCh38
rs200326674 Chr.1: 3858750-3858750 on Build GRCh38
rs752601088 Chr.1: 3858762-3858762 on Build GRCh38
rs770254314 Chr.1: 3858688-3858688 on Build GRCh38
rs781238042 Chr.1: 3858765-3858765 on Build GRCh38
rs761446656 Chr.1: 3858712-3858712 on Build GRCh38
rs781435042 Chr.1: 3858683-3858683 on Build GRCh38
rs746087211 Chr.1: 3858686-3858686 on Build GRCh38
rs891046829 Chr.1: 3858843-3858843 on Build GRCh38
rs201900322 Chr.1: 3858770-3858770 on Build GRCh38
rs771700224 Chr.1: 3858728-3858728 on Build GRCh38
rs770441698 Chr.1: 3858821-3858821 on Build GRCh38
rs370680894 Chr.1: 3858730-3858730 on Build GRCh38
rs1051493287 Chr.1: 3858816-3858816 on Build GRCh38
rs757305966 Chr.1: 3858673-3858673 on Build GRCh38
rs748649635 Chr.1: 3858782-3858782 on Build GRCh38
rs781186974 Chr.1: 3858719-3858719 on Build GRCh38
rs765014861 Chr.1: 3858838-3858838 on Build GRCh38
rs528823646 Chr.1: 3858752-3858752 on Build GRCh38
rs148359074 Chr.1: 3858708-3858708 on Build GRCh38
rs748592254 Chr.1: 3858681-3858681 on Build GRCh38
rs1009173674 Chr.1: 3858649-3858649 on Build GRCh38

View Full Product Details