Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 150221482-150221753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK2
Entrez Gene ID
166614
Gene Name
doublecortin like kinase 2
Gene Aliases
CL2, CLICK-II, CLICK2, CLIK2, DCAMKL2, DCDC3, DCDC3B, DCK2
UniGene
Hs.591683
Gene Chromosome Location
Chr.4: 150078274-150258563 on Build GRCh38
Transcripts
NM_001040260.3 NM_001040261.4 NR_036614.1 XM_005262785.2 XM_005262786.2 XM_017007832.1 XM_017007833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5692881 c.1174G>C p.E392Q Chr.4: 150221718-150221718 on Build GRCh38
7110060 c.1133-1G>T p.? Chr.4: 150221676-150221676 on Build GRCh38
5687960 c.1205C>G p.S402C Chr.4: 150221698-150221698 on Build GRCh38
1051946 c.1193A>T p.K398M Chr.4: 150221737-150221737 on Build GRCh38
5692880 c.1225G>C p.E409Q Chr.4: 150221718-150221718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369416111 Chr.4: 150221670-150221670 on Build GRCh38
rs143364310 Chr.4: 150221552-150221552 on Build GRCh38
rs758538840 Chr.4: 150221700-150221700 on Build GRCh38
rs911998614 Chr.4: 150221630-150221630 on Build GRCh38
rs550153471 Chr.4: 150221594-150221594 on Build GRCh38
rs142061527 Chr.4: 150221701-150221701 on Build GRCh38
rs972235763 Chr.4: 150221672-150221672 on Build GRCh38
rs986644037 Chr.4: 150221617-150221617 on Build GRCh38
rs773057815 Chr.4: 150221712-150221712 on Build GRCh38
rs998655527 Chr.4: 150221568-150221568 on Build GRCh38
rs552557131 Chr.4: 150221710-150221710 on Build GRCh38
rs561454262 Chr.4: 150221563-150221563 on Build GRCh38
rs369090070 Chr.4: 150221626-150221627 on Build GRCh38
rs766901912 Chr.4: 150221604-150221604 on Build GRCh38
rs372651298 Chr.4: 150221705-150221705 on Build GRCh38
rs746315066 Chr.4: 150221696-150221696 on Build GRCh38
rs182654859 Chr.4: 150221583-150221583 on Build GRCh38
rs1049208625 Chr.4: 150221510-150221510 on Build GRCh38
rs930923607 Chr.4: 150221693-150221693 on Build GRCh38
rs957286423 Chr.4: 150221589-150221589 on Build GRCh38
rs558928288 Chr.4: 150221540-150221540 on Build GRCh38
rs757748738 Chr.4: 150221673-150221673 on Build GRCh38
rs931266664 Chr.4: 150221516-150221516 on Build GRCh38
rs571606085 Chr.4: 150221627-150221627 on Build GRCh38
rs764563049 Chr.4: 150221640-150221640 on Build GRCh38
rs769739787 Chr.4: 150221592-150221592 on Build GRCh38
rs576691132 Chr.4: 150221512-150221512 on Build GRCh38
rs779456964 Chr.4: 150221686-150221686 on Build GRCh38
rs527948845 Chr.4: 150221550-150221550 on Build GRCh38
rs754238415 Chr.4: 150221642-150221642 on Build GRCh38
rs201098370 Chr.4: 150221635-150221635 on Build GRCh38
rs72951529 Chr.4: 150221596-150221596 on Build GRCh38
rs745911245 Chr.4: 150221588-150221588 on Build GRCh38
rs879654853 Chr.4: 150221532-150221532 on Build GRCh38
rs113059899 Chr.4: 150221709-150221709 on Build GRCh38

View Full Product Details