Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52656761-52657012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM159A
Entrez Gene ID
348378
Gene Name
family with sequence similarity 159 member A
Gene Aliases
PRO7171, WWLS2783
UniGene
Hs.424589
Gene Chromosome Location
Chr.1: 52631438-52669120 on Build GRCh38
Transcripts
NM_001042693.2 NR_135156.1 NR_135157.1 XM_011541377.2 XM_011541378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4188786 c.538G>C p.V180L Chr.1: 52657005-52657005 on Build GRCh38
4008833 c.470T>C p.L157P Chr.1: 52656937-52656937 on Build GRCh38
4008832 c.438C>T p.S146S Chr.1: 52656905-52656905 on Build GRCh38
6658698 c.467G>A p.R156H Chr.1: 52656934-52656934 on Build GRCh38
6540449 c.379G>T p.V127L Chr.1: 52656846-52656846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780102564 Chr.1: 52656961-52656961 on Build GRCh38
rs756846089 Chr.1: 52656846-52656846 on Build GRCh38
rs746634577 Chr.1: 52656985-52656987 on Build GRCh38
rs778859654 Chr.1: 52656980-52656980 on Build GRCh38
rs372921320 Chr.1: 52656899-52656899 on Build GRCh38
rs753703125 Chr.1: 52656966-52656966 on Build GRCh38
rs774402161 Chr.1: 52656801-52656801 on Build GRCh38
rs761268351 Chr.1: 52656828-52656828 on Build GRCh38
rs372989522 Chr.1: 52656957-52656957 on Build GRCh38
rs762583405 Chr.1: 52656936-52656936 on Build GRCh38
rs376662330 Chr.1: 52656841-52656841 on Build GRCh38
rs761487827 Chr.1: 52656950-52656950 on Build GRCh38
rs772546895 Chr.1: 52656817-52656817 on Build GRCh38
rs751004636 Chr.1: 52656946-52656946 on Build GRCh38
rs369065271 Chr.1: 52656845-52656845 on Build GRCh38
rs201585698 Chr.1: 52656933-52656933 on Build GRCh38
rs1016718670 Chr.1: 52656791-52656791 on Build GRCh38
rs749280895 Chr.1: 52656795-52656795 on Build GRCh38
rs768974033 Chr.1: 52656800-52656800 on Build GRCh38
rs577780949 Chr.1: 52656927-52656927 on Build GRCh38
rs372709261 Chr.1: 52656836-52656836 on Build GRCh38
rs748392048 Chr.1: 52656911-52656911 on Build GRCh38
rs780217057 Chr.1: 52656781-52656781 on Build GRCh38
rs761070144 Chr.1: 52656923-52656923 on Build GRCh38
rs145105252 Chr.1: 52656976-52656976 on Build GRCh38
rs1010540296 Chr.1: 52656824-52656824 on Build GRCh38
rs779249380 Chr.1: 52656905-52656905 on Build GRCh38
rs977394483 Chr.1: 52656851-52656851 on Build GRCh38
rs756070190 Chr.1: 52656872-52656872 on Build GRCh38
rs752273057 Chr.1: 52656838-52656838 on Build GRCh38
rs768612287 Chr.1: 52656902-52656902 on Build GRCh38
rs749422484 Chr.1: 52656893-52656893 on Build GRCh38
rs201777412 Chr.1: 52656918-52656918 on Build GRCh38
rs764538952 Chr.1: 52656860-52656860 on Build GRCh38
rs763525572 Chr.1: 52656943-52656943 on Build GRCh38
rs200338087 Chr.1: 52656913-52656913 on Build GRCh38
rs777001067 Chr.1: 52656934-52656934 on Build GRCh38
rs767112210 Chr.1: 52656956-52656956 on Build GRCh38
rs755875754 Chr.1: 52656959-52656959 on Build GRCh38
rs762333098 Chr.1: 52656813-52656813 on Build GRCh38
rs762346974 Chr.1: 52656839-52656839 on Build GRCh38
rs779890334 Chr.1: 52656961-52656970 on Build GRCh38
rs773649586 Chr.1: 52656820-52656820 on Build GRCh38
rs773743354 Chr.1: 52656914-52656914 on Build GRCh38
rs779836276 Chr.1: 52656892-52656892 on Build GRCh38
rs750182514 Chr.1: 52656854-52656854 on Build GRCh38
rs895355538 Chr.1: 52656944-52656944 on Build GRCh38
rs763348426 Chr.1: 52656807-52656807 on Build GRCh38

View Full Product Details