Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121813821-121814068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC9
Entrez Gene ID
5393
Gene Name
exosome component 9
Gene Aliases
PM/Scl-75, PMSCL1, RRP45, Rrp45p, p5, p6
UniGene
Hs.91728
Gene Chromosome Location
Chr.4: 121801295-121817021 on Build GRCh38
Transcripts
NM_001034194.1 NM_005033.2 XM_011532035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4515258 c.1010_1011CC>TT p.A337V Chr.4: 121813901-121813902 on Build GRCh38
7134627 c.1096A>G p.I366V Chr.4: 121813987-121813987 on Build GRCh38
7274415 c.993A>G p.L331L Chr.4: 121813884-121813884 on Build GRCh38
5440081 c.1043G>T p.G348V Chr.4: 121813934-121813934 on Build GRCh38
6345872 c.1109G>T p.G370V Chr.4: 121814000-121814000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546850964 Chr.4: 121813891-121813891 on Build GRCh38
rs186660763 Chr.4: 121813858-121813858 on Build GRCh38
rs777127703 Chr.4: 121813936-121813936 on Build GRCh38
rs761031481 Chr.4: 121813875-121813875 on Build GRCh38
rs112371984 Chr.4: 121813865-121813865 on Build GRCh38
rs76155182 Chr.4: 121813977-121813977 on Build GRCh38
rs745817801 Chr.4: 121813955-121813955 on Build GRCh38
rs778119795 Chr.4: 121813876-121813876 on Build GRCh38
rs762201983 Chr.4: 121813905-121813905 on Build GRCh38
rs935086657 Chr.4: 121814031-121814031 on Build GRCh38
rs761911321 Chr.4: 121813983-121813983 on Build GRCh38
rs750618265 Chr.4: 121813947-121813948 on Build GRCh38
rs142828776 Chr.4: 121813883-121813883 on Build GRCh38
rs1803183 Chr.4: 121813987-121813987 on Build GRCh38
rs766699400 Chr.4: 121814027-121814027 on Build GRCh38
rs938448213 Chr.4: 121813986-121813987 on Build GRCh38
rs564581752 Chr.4: 121813884-121813884 on Build GRCh38
rs755322508 Chr.4: 121814037-121814037 on Build GRCh38
rs762048513 Chr.4: 121813889-121813890 on Build GRCh38
rs751970914 Chr.4: 121814036-121814036 on Build GRCh38
rs963357147 Chr.4: 121813925-121813925 on Build GRCh38
rs1019478137 Chr.4: 121813879-121813879 on Build GRCh38
rs768969604 Chr.4: 121813973-121813973 on Build GRCh38
rs778847636 Chr.4: 121813948-121813948 on Build GRCh38
rs765642150 Chr.4: 121813917-121813917 on Build GRCh38
rs748785283 Chr.4: 121813939-121813939 on Build GRCh38
rs752343205 Chr.4: 121813928-121813928 on Build GRCh38
rs982858939 Chr.4: 121813976-121813976 on Build GRCh38
rs148293459 Chr.4: 121813972-121813972 on Build GRCh38
rs763257543 Chr.4: 121813993-121813994 on Build GRCh38
rs763519680 Chr.4: 121814008-121814008 on Build GRCh38
rs756751733 Chr.4: 121813941-121813941 on Build GRCh38
rs1030461107 Chr.4: 121813856-121813856 on Build GRCh38
rs143249010 Chr.4: 121813971-121813971 on Build GRCh38
rs963898844 Chr.4: 121813890-121813890 on Build GRCh38
rs542804602 Chr.4: 121813910-121813910 on Build GRCh38
rs758695368 Chr.4: 121813923-121813923 on Build GRCh38
rs201123592 Chr.4: 121813924-121813924 on Build GRCh38
rs745621681 Chr.4: 121813857-121813857 on Build GRCh38
rs533678536 Chr.4: 121813888-121813888 on Build GRCh38
rs771975787 Chr.4: 121813962-121813962 on Build GRCh38
rs374466564 Chr.4: 121813873-121813873 on Build GRCh38
rs772000650 Chr.4: 121813870-121813870 on Build GRCh38
rs768578666 Chr.4: 121813984-121813984 on Build GRCh38
rs755805732 Chr.4: 121813929-121813929 on Build GRCh38
rs113512885 Chr.4: 121813864-121813864 on Build GRCh38

View Full Product Details