Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 990137-990388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT3
Entrez Gene ID
58524
Gene Name
doublesex and mab-3 related transcription factor 3
Gene Aliases
DMRTA3
UniGene
Hs.189174
Gene Chromosome Location
Chr.9: 976968-991732 on Build GRCh38
Transcripts
NM_021240.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310621 c.232G>T p.E78* Chr.9: 990229-990229 on Build GRCh38
1728662 c.731G>T p.S244I Chr.9: 990317-990317 on Build GRCh38
2733599 c.232G>A p.E78K Chr.9: 990229-990229 on Build GRCh38
4394466 c.690C>T p.L230L Chr.9: 990276-990276 on Build GRCh38
70469 c.761T>A p.V254E Chr.9: 990347-990347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539407759 Chr.9: 990183-990183 on Build GRCh38
rs150599487 Chr.9: 990335-990335 on Build GRCh38
rs570625531 Chr.9: 990165-990165 on Build GRCh38
rs752829841 Chr.9: 990202-990202 on Build GRCh38
rs766278011 Chr.9: 990201-990201 on Build GRCh38
rs749745516 Chr.9: 990229-990229 on Build GRCh38
rs778528769 Chr.9: 990291-990291 on Build GRCh38
rs139342042 Chr.9: 990309-990309 on Build GRCh38
rs775178489 Chr.9: 990163-990163 on Build GRCh38
rs796689543 Chr.9: 990222-990222 on Build GRCh38
rs779277340 Chr.9: 990365-990365 on Build GRCh38
rs777202996 Chr.9: 990369-990369 on Build GRCh38
rs538464950 Chr.9: 990234-990234 on Build GRCh38
rs762281629 Chr.9: 990193-990193 on Build GRCh38
rs569515243 Chr.9: 990190-990190 on Build GRCh38
rs555913389 Chr.9: 990227-990227 on Build GRCh38
rs757372153 Chr.9: 990215-990215 on Build GRCh38
rs144138014 Chr.9: 990330-990330 on Build GRCh38
rs202197205 Chr.9: 990228-990228 on Build GRCh38
rs747592507 Chr.9: 990297-990297 on Build GRCh38
rs772844218 Chr.9: 990305-990305 on Build GRCh38
rs559135481 Chr.9: 990316-990316 on Build GRCh38
rs780509910 Chr.9: 990275-990275 on Build GRCh38
rs372697831 Chr.9: 990338-990338 on Build GRCh38
rs77818517 Chr.9: 990258-990258 on Build GRCh38
rs781642811 Chr.9: 990216-990216 on Build GRCh38
rs986374173 Chr.9: 990159-990159 on Build GRCh38
rs957559381 Chr.9: 990327-990327 on Build GRCh38
rs184249595 Chr.9: 990339-990339 on Build GRCh38
rs567149898 Chr.9: 990204-990204 on Build GRCh38
rs780127464 Chr.9: 990357-990357 on Build GRCh38
rs376961643 Chr.9: 990345-990345 on Build GRCh38
rs149469082 Chr.9: 990184-990184 on Build GRCh38
rs1014143151 Chr.9: 990294-990294 on Build GRCh38
rs773043267 Chr.9: 990236-990236 on Build GRCh38
rs751707746 Chr.9: 990326-990326 on Build GRCh38
rs199530929 Chr.9: 990336-990336 on Build GRCh38
rs752903154 Chr.9: 990260-990260 on Build GRCh38
rs1005377434 Chr.9: 990243-990243 on Build GRCh38
rs61737966 Chr.9: 990235-990235 on Build GRCh38
rs7854621 Chr.9: 990368-990368 on Build GRCh38
rs374084588 Chr.9: 990158-990158 on Build GRCh38
rs143922008 Chr.9: 990212-990212 on Build GRCh38
rs139232109 Chr.9: 990356-990356 on Build GRCh38
rs759330661 Chr.9: 990249-990249 on Build GRCh38
rs749839281 Chr.9: 990288-990288 on Build GRCh38
rs367997053 Chr.9: 990213-990213 on Build GRCh38
rs757538425 Chr.9: 990259-990259 on Build GRCh38
rs766367767 Chr.9: 990238-990238 on Build GRCh38
rs535812289 Chr.9: 990217-990217 on Build GRCh38
rs575777699 Chr.9: 990233-990233 on Build GRCh38
rs955958093 Chr.9: 990265-990265 on Build GRCh38
rs894388369 Chr.9: 990266-990266 on Build GRCh38
rs765052304 Chr.9: 990317-990317 on Build GRCh38
rs750182111 Chr.9: 990189-990189 on Build GRCh38
rs776360310 Chr.9: 990310-990310 on Build GRCh38
rs771811953 Chr.9: 990302-990302 on Build GRCh38
rs769008836 Chr.9: 990290-990290 on Build GRCh38
rs753776043 Chr.9: 990239-990239 on Build GRCh38
rs763720874 Chr.9: 990167-990167 on Build GRCh38
rs774058341 Chr.9: 990176-990176 on Build GRCh38
rs756554759 Chr.9: 990263-990263 on Build GRCh38
rs760447078 Chr.9: 990237-990237 on Build GRCh38
rs376074221 Chr.9: 990198-990198 on Build GRCh38
rs148221558 Chr.9: 990221-990221 on Build GRCh38
rs1031623571 Chr.9: 990255-990255 on Build GRCh38

View Full Product Details