Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89178833-89179092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR1
Entrez Gene ID
2569
Gene Name
gamma-aminobutyric acid type A receptor rho1 subunit
Gene Aliases
-
UniGene
Hs.99927
Gene Chromosome Location
Chr.6: 89177504-89231288 on Build GRCh38
Transcripts
NM_001256703.1 NM_001256704.1 NM_001267582.1 NM_002042.4 XM_017010689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668722 c.1338G>A p.V446V Chr.6: 89178872-89178872 on Build GRCh38
3674761 c.1324A>C p.R442R Chr.6: 89178886-89178886 on Build GRCh38
1643164 c.1177C>T p.R393C Chr.6: 89179033-89179033 on Build GRCh38
363415 c.1285C>A p.L429M Chr.6: 89178925-89178925 on Build GRCh38
5886470 c.1151C>T p.P384L Chr.6: 89179059-89179059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375411182 Chr.6: 89178907-89178907 on Build GRCh38
rs201665605 Chr.6: 89179011-89179011 on Build GRCh38
rs772321848 Chr.6: 89178937-89178937 on Build GRCh38
rs1796743 Chr.6: 89179025-89179025 on Build GRCh38
rs779365894 Chr.6: 89178927-89178927 on Build GRCh38
rs757804410 Chr.6: 89178974-89178974 on Build GRCh38
rs200078707 Chr.6: 89178957-89178957 on Build GRCh38
rs146705571 Chr.6: 89179060-89179060 on Build GRCh38
rs780193466 Chr.6: 89178990-89178990 on Build GRCh38
rs755625493 Chr.6: 89178959-89178959 on Build GRCh38
rs777339555 Chr.6: 89178963-89178963 on Build GRCh38
rs749813743 Chr.6: 89178924-89178924 on Build GRCh38
rs539734627 Chr.6: 89178998-89178998 on Build GRCh38
rs757324147 Chr.6: 89179068-89179068 on Build GRCh38
rs143260597 Chr.6: 89179059-89179059 on Build GRCh38
rs766203740 Chr.6: 89178872-89178872 on Build GRCh38
rs773931969 Chr.6: 89179034-89179035 on Build GRCh38
rs752292472 Chr.6: 89178902-89178902 on Build GRCh38
rs530003842 Chr.6: 89179016-89179016 on Build GRCh38
rs772958770 Chr.6: 89178948-89178948 on Build GRCh38
rs750943576 Chr.6: 89179042-89179042 on Build GRCh38
rs373658230 Chr.6: 89178875-89178875 on Build GRCh38
rs375182799 Chr.6: 89179038-89179038 on Build GRCh38
rs545780993 Chr.6: 89179052-89179052 on Build GRCh38
rs549063825 Chr.6: 89178934-89178934 on Build GRCh38
rs370391229 Chr.6: 89179037-89179037 on Build GRCh38
rs760804771 Chr.6: 89178939-89178939 on Build GRCh38
rs775545112 Chr.6: 89178938-89178938 on Build GRCh38
rs867047200 Chr.6: 89178899-89178899 on Build GRCh38
rs768738423 Chr.6: 89178940-89178940 on Build GRCh38
rs202214989 Chr.6: 89179026-89179026 on Build GRCh38
rs776799073 Chr.6: 89179014-89179014 on Build GRCh38
rs372823219 Chr.6: 89179001-89179001 on Build GRCh38
rs912306660 Chr.6: 89179048-89179048 on Build GRCh38
rs200988696 Chr.6: 89178949-89178949 on Build GRCh38
rs754704522 Chr.6: 89178885-89178885 on Build GRCh38
rs770188872 Chr.6: 89178987-89178987 on Build GRCh38
rs780822875 Chr.6: 89178889-89178889 on Build GRCh38
rs118018352 Chr.6: 89179033-89179033 on Build GRCh38
rs767341814 Chr.6: 89178955-89178955 on Build GRCh38
rs147082491 Chr.6: 89178989-89178989 on Build GRCh38
rs762600089 Chr.6: 89178862-89178862 on Build GRCh38
rs969481270 Chr.6: 89178970-89178970 on Build GRCh38
rs757205613 Chr.6: 89178973-89178973 on Build GRCh38
rs750220329 Chr.6: 89178860-89178860 on Build GRCh38
rs779117020 Chr.6: 89178980-89178980 on Build GRCh38
rs201104884 Chr.6: 89179027-89179027 on Build GRCh38
rs765227482 Chr.6: 89178859-89178859 on Build GRCh38
rs370239040 Chr.6: 89179032-89179032 on Build GRCh38
rs767431536 Chr.6: 89179028-89179028 on Build GRCh38
rs749163278 Chr.6: 89179015-89179015 on Build GRCh38
rs373529328 Chr.6: 89178960-89178960 on Build GRCh38
rs368507511 Chr.6: 89179066-89179066 on Build GRCh38
rs753894820 Chr.6: 89178892-89178892 on Build GRCh38
rs778199318 Chr.6: 89178915-89178915 on Build GRCh38
rs771132769 Chr.6: 89178925-89178925 on Build GRCh38
rs908600319 Chr.6: 89179020-89179020 on Build GRCh38
rs775214942 Chr.6: 89179030-89179030 on Build GRCh38
rs143144903 Chr.6: 89178950-89178950 on Build GRCh38

View Full Product Details