Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20042320-20042569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
UniGene
Hs.171342
Gene Chromosome Location
Chr.20: 20034361-20056046 on Build GRCh38
Transcripts
NM_001278625.1 NM_001278626.1 NM_001278627.1 NM_001278628.1 NM_016652.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246311 c.1478C>A p.A493E Chr.20: 20042494-20042494 on Build GRCh38
4289971 c.1518A>C p.E506D Chr.20: 20042454-20042454 on Build GRCh38
160213 c.1632A>G p.E544E Chr.20: 20042340-20042340 on Build GRCh38
5968003 c.1557C>T p.A519A Chr.20: 20042415-20042415 on Build GRCh38
6017820 c.1526C>T p.A509V Chr.20: 20042446-20042446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146936626 Chr.20: 20042476-20042476 on Build GRCh38
rs748004236 Chr.20: 20042429-20042430 on Build GRCh38
rs577483196 Chr.20: 20042477-20042477 on Build GRCh38
rs3828004 Chr.20: 20042530-20042530 on Build GRCh38
rs759306938 Chr.20: 20042403-20042403 on Build GRCh38
rs1026870452 Chr.20: 20042504-20042504 on Build GRCh38
rs777276034 Chr.20: 20042388-20042388 on Build GRCh38
rs778762710 Chr.20: 20042459-20042459 on Build GRCh38
rs755388127 Chr.20: 20042446-20042446 on Build GRCh38
rs777262990 Chr.20: 20042542-20042542 on Build GRCh38
rs745732279 Chr.20: 20042479-20042479 on Build GRCh38
rs141192187 Chr.20: 20042472-20042472 on Build GRCh38
rs750451055 Chr.20: 20042431-20042431 on Build GRCh38
rs777183138 Chr.20: 20042462-20042462 on Build GRCh38
rs757665657 Chr.20: 20042411-20042411 on Build GRCh38
rs1035672020 Chr.20: 20042366-20042366 on Build GRCh38
rs371056596 Chr.20: 20042505-20042505 on Build GRCh38
rs370539789 Chr.20: 20042392-20042392 on Build GRCh38
rs767508370 Chr.20: 20042413-20042413 on Build GRCh38
rs746921866 Chr.20: 20042389-20042389 on Build GRCh38
rs553862251 Chr.20: 20042373-20042373 on Build GRCh38
rs753567981 Chr.20: 20042352-20042352 on Build GRCh38
rs776317200 Chr.20: 20042400-20042400 on Build GRCh38
rs771691914 Chr.20: 20042387-20042387 on Build GRCh38
rs1025704944 Chr.20: 20042499-20042499 on Build GRCh38
rs775209312 Chr.20: 20042493-20042493 on Build GRCh38
rs768909768 Chr.20: 20042409-20042409 on Build GRCh38
rs752407308 Chr.20: 20042460-20042460 on Build GRCh38
rs761108473 Chr.20: 20042437-20042437 on Build GRCh38
rs766860396 Chr.20: 20042538-20042538 on Build GRCh38
rs766886257 Chr.20: 20042442-20042442 on Build GRCh38
rs773213828 Chr.20: 20042528-20042528 on Build GRCh38
rs1015834536 Chr.20: 20042454-20042454 on Build GRCh38
rs778674808 Chr.20: 20042368-20042368 on Build GRCh38
rs772288207 Chr.20: 20042514-20042514 on Build GRCh38
rs146257632 Chr.20: 20042445-20042445 on Build GRCh38
rs754725111 Chr.20: 20042353-20042353 on Build GRCh38
rs769565084 Chr.20: 20042492-20042492 on Build GRCh38
rs1001789559 Chr.20: 20042415-20042415 on Build GRCh38

View Full Product Details