Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154002200-154002471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A2
Entrez Gene ID
10827
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
133K02, C5orf3
UniGene
Hs.166551
Gene Chromosome Location
Chr.5: 153990131-154038938 on Build GRCh38
Transcripts
NM_001317993.1 NM_001317994.1 NM_001317995.1 NM_018691.3 XM_005268360.4 XM_005268361.4 XM_011537540.2 XM_011537541.2 XM_011537542.2 XM_017008967.1 XM_017008968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160883 c.1179C>A p.F393L Chr.5: 154002328-154002328 on Build GRCh38
7144756 c.1155G>C p.L385L Chr.5: 154002352-154002352 on Build GRCh38
7144755 c.1239G>C p.R413S Chr.5: 154002268-154002268 on Build GRCh38
5697182 c.1155G>A p.L385L Chr.5: 154002352-154002352 on Build GRCh38
4762878 c.1133C>T p.A378V Chr.5: 154002374-154002374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374460608 Chr.5: 154002376-154002376 on Build GRCh38
rs918599750 Chr.5: 154002440-154002440 on Build GRCh38
rs767630433 Chr.5: 154002260-154002260 on Build GRCh38
rs766099452 Chr.5: 154002379-154002379 on Build GRCh38
rs199742756 Chr.5: 154002262-154002262 on Build GRCh38
rs773665749 Chr.5: 154002246-154002246 on Build GRCh38
rs775588851 Chr.5: 154002365-154002365 on Build GRCh38
rs778482580 Chr.5: 154002431-154002431 on Build GRCh38
rs758154392 Chr.5: 154002278-154002278 on Build GRCh38
rs761767529 Chr.5: 154002368-154002368 on Build GRCh38
rs201640396 Chr.5: 154002258-154002258 on Build GRCh38
rs1016436748 Chr.5: 154002322-154002322 on Build GRCh38
rs973708154 Chr.5: 154002389-154002389 on Build GRCh38
rs761041894 Chr.5: 154002361-154002361 on Build GRCh38
rs201795266 Chr.5: 154002282-154002282 on Build GRCh38
rs370862982 Chr.5: 154002370-154002370 on Build GRCh38
rs749806938 Chr.5: 154002432-154002432 on Build GRCh38
rs780697366 Chr.5: 154002301-154002301 on Build GRCh38
rs753721633 Chr.5: 154002261-154002261 on Build GRCh38
rs1017094618 Chr.5: 154002404-154002405 on Build GRCh38
rs11748836 Chr.5: 154002288-154002288 on Build GRCh38
rs74196375 Chr.5: 154002438-154002438 on Build GRCh38
rs554683102 Chr.5: 154002251-154002251 on Build GRCh38
rs760462959 Chr.5: 154002356-154002371 on Build GRCh38
rs766830399 Chr.5: 154002395-154002395 on Build GRCh38
rs766970633 Chr.5: 154002396-154002396 on Build GRCh38
rs183682516 Chr.5: 154002425-154002425 on Build GRCh38
rs761751709 Chr.5: 154002270-154002270 on Build GRCh38
rs766334340 Chr.5: 154002376-154002376 on Build GRCh38
rs747024457 Chr.5: 154002241-154002246 on Build GRCh38
rs775272637 Chr.5: 154002380-154002380 on Build GRCh38
rs953856114 Chr.5: 154002424-154002424 on Build GRCh38
rs750176286 Chr.5: 154002276-154002276 on Build GRCh38
rs754382121 Chr.5: 154002405-154002405 on Build GRCh38
rs754454894 Chr.5: 154002292-154002292 on Build GRCh38
rs774503196 Chr.5: 154002345-154002345 on Build GRCh38
rs765196839 Chr.5: 154002369-154002369 on Build GRCh38
rs138263333 Chr.5: 154002313-154002313 on Build GRCh38
rs753922199 Chr.5: 154002438-154002441 on Build GRCh38
rs774611281 Chr.5: 154002253-154002253 on Build GRCh38
rs771409700 Chr.5: 154002339-154002339 on Build GRCh38
rs545649031 Chr.5: 154002387-154002387 on Build GRCh38
rs778396391 Chr.5: 154002323-154002323 on Build GRCh38
rs748625813 Chr.5: 154002242-154002242 on Build GRCh38
rs773106930 Chr.5: 154002372-154002372 on Build GRCh38
rs749719044 Chr.5: 154002324-154002324 on Build GRCh38
rs772056516 Chr.5: 154002360-154002360 on Build GRCh38
rs767864959 Chr.5: 154002244-154002244 on Build GRCh38
rs1002989258 Chr.5: 154002255-154002255 on Build GRCh38
rs746035433 Chr.5: 154002355-154002355 on Build GRCh38
rs763300211 Chr.5: 154002247-154002247 on Build GRCh38

View Full Product Details