Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72012089-72012336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
UniGene
Hs.654427
Gene Chromosome Location
Chr.16: 72008744-72027659 on Build GRCh38
Transcripts
NM_001361.4 XM_005255827.3 XM_017022990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4262205 c.178C>T p.R60C Chr.16: 72012206-72012206 on Build GRCh38
6643285 c.93G>A p.T31T Chr.16: 72012121-72012121 on Build GRCh38
4757270 c.103C>T p.R35C Chr.16: 72012131-72012131 on Build GRCh38
4062625 c.67C>A p.L23I Chr.16: 72012095-72012095 on Build GRCh38
1203513 c.92C>T p.T31M Chr.16: 72012120-72012120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748091623 Chr.16: 72012228-72012228 on Build GRCh38
rs375281379 Chr.16: 72012207-72012207 on Build GRCh38
rs373700454 Chr.16: 72012130-72012130 on Build GRCh38
rs749041532 Chr.16: 72012217-72012217 on Build GRCh38
rs745891169 Chr.16: 72012257-72012257 on Build GRCh38
rs372155275 Chr.16: 72012180-72012180 on Build GRCh38
rs942178508 Chr.16: 72012150-72012150 on Build GRCh38
rs776827112 Chr.16: 72012234-72012234 on Build GRCh38
rs368395651 Chr.16: 72012182-72012182 on Build GRCh38
rs779682949 Chr.16: 72012121-72012121 on Build GRCh38
rs769053767 Chr.16: 72012271-72012271 on Build GRCh38
rs189332001 Chr.16: 72012204-72012204 on Build GRCh38
rs779717056 Chr.16: 72012213-72012213 on Build GRCh38
rs752050272 Chr.16: 72012199-72012199 on Build GRCh38
rs763244296 Chr.16: 72012268-72012268 on Build GRCh38
rs778735330 Chr.16: 72012309-72012309 on Build GRCh38
rs755971761 Chr.16: 72012120-72012120 on Build GRCh38
rs377442561 Chr.16: 72012305-72012305 on Build GRCh38
rs767063416 Chr.16: 72012286-72012286 on Build GRCh38
rs1015555124 Chr.16: 72012110-72012110 on Build GRCh38
rs745682638 Chr.16: 72012159-72012160 on Build GRCh38
rs8057016 Chr.16: 72012312-72012312 on Build GRCh38
rs368757082 Chr.16: 72012131-72012131 on Build GRCh38
rs147756052 Chr.16: 72012202-72012203 on Build GRCh38
rs754794207 Chr.16: 72012306-72012306 on Build GRCh38
rs936867357 Chr.16: 72012214-72012214 on Build GRCh38
rs777056927 Chr.16: 72012157-72012157 on Build GRCh38
rs765741816 Chr.16: 72012172-72012172 on Build GRCh38
rs77625181 Chr.16: 72012301-72012301 on Build GRCh38
rs370028170 Chr.16: 72012287-72012287 on Build GRCh38
rs972083429 Chr.16: 72012139-72012139 on Build GRCh38
rs375174980 Chr.16: 72012168-72012168 on Build GRCh38
rs931958752 Chr.16: 72012206-72012206 on Build GRCh38
rs199675109 Chr.16: 72012132-72012132 on Build GRCh38
rs945616673 Chr.16: 72012250-72012251 on Build GRCh38
rs750116974 Chr.16: 72012209-72012209 on Build GRCh38
rs369448475 Chr.16: 72012220-72012220 on Build GRCh38
rs752259247 Chr.16: 72012191-72012192 on Build GRCh38
rs979830995 Chr.16: 72012112-72012112 on Build GRCh38
rs375435688 Chr.16: 72012181-72012181 on Build GRCh38
rs778771847 Chr.16: 72012224-72012224 on Build GRCh38
rs113393105 Chr.16: 72012302-72012302 on Build GRCh38
rs200812275 Chr.16: 72012283-72012283 on Build GRCh38
rs775719313 Chr.16: 72012266-72012266 on Build GRCh38
rs770175382 Chr.16: 72012265-72012265 on Build GRCh38
rs747071335 Chr.16: 72012151-72012151 on Build GRCh38
rs201823503 Chr.16: 72012233-72012233 on Build GRCh38
rs200842559 Chr.16: 72012156-72012156 on Build GRCh38
rs370288406 Chr.16: 72012118-72012118 on Build GRCh38
rs755742397 Chr.16: 72012211-72012211 on Build GRCh38

View Full Product Details