Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82252899-82253160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1D
Entrez Gene ID
1453
Gene Name
casein kinase 1 delta
Gene Aliases
ASPS, CKIdelta, FASPS2, HCKID
UniGene
Hs.631725
Gene Chromosome Location
Chr.17: 82239317-82273742 on Build GRCh38
Transcripts
NM_001893.4 NM_139062.2 NR_110578.1 XM_005256336.3 XM_005256337.4 XM_017024199.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242163 c.451G>T p.G151W Chr.17: 82253130-82253130 on Build GRCh38
5498293 c.453G>A p.G151G Chr.17: 82253128-82253128 on Build GRCh38
4403044 c.444C>G p.I148M Chr.17: 82253137-82253137 on Build GRCh38
1387463 c.537C>T p.Y179Y Chr.17: 82253044-82253044 on Build GRCh38
6636732 c.479G>A p.R160H Chr.17: 82253102-82253102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900928787 Chr.17: 82252953-82252953 on Build GRCh38
rs542306409 Chr.17: 82252938-82252938 on Build GRCh38
rs1033299165 Chr.17: 82252966-82252966 on Build GRCh38
rs980797953 Chr.17: 82252931-82252931 on Build GRCh38
rs769584405 Chr.17: 82253007-82253007 on Build GRCh38
rs748668818 Chr.17: 82252996-82252996 on Build GRCh38
rs146479130 Chr.17: 82253044-82253044 on Build GRCh38
rs756358012 Chr.17: 82252979-82252979 on Build GRCh38
rs770044849 Chr.17: 82252934-82252934 on Build GRCh38
rs373197686 Chr.17: 82252988-82252988 on Build GRCh38
rs888400777 Chr.17: 82253099-82253099 on Build GRCh38
rs201710093 Chr.17: 82253131-82253131 on Build GRCh38
rs771661652 Chr.17: 82253107-82253107 on Build GRCh38
rs750648327 Chr.17: 82252971-82252971 on Build GRCh38
rs762907794 Chr.17: 82253043-82253043 on Build GRCh38
rs978987880 Chr.17: 82252964-82252964 on Build GRCh38
rs189567680 Chr.17: 82252955-82252955 on Build GRCh38
rs953764648 Chr.17: 82252961-82252961 on Build GRCh38
rs144919619 Chr.17: 82253104-82253104 on Build GRCh38
rs761776300 Chr.17: 82253059-82253059 on Build GRCh38
rs780197149 Chr.17: 82252983-82252983 on Build GRCh38
rs749662977 Chr.17: 82252987-82252987 on Build GRCh38
rs138788002 Chr.17: 82252990-82252990 on Build GRCh38
rs775217166 Chr.17: 82253029-82253029 on Build GRCh38
rs572190875 Chr.17: 82252956-82252956 on Build GRCh38
rs1004041864 Chr.17: 82252933-82252933 on Build GRCh38
rs946815652 Chr.17: 82252923-82252923 on Build GRCh38
rs369895747 Chr.17: 82252969-82252969 on Build GRCh38
rs766658391 Chr.17: 82253112-82253112 on Build GRCh38
rs140988157 Chr.17: 82253074-82253074 on Build GRCh38
rs369286452 Chr.17: 82253000-82253000 on Build GRCh38
rs557207079 Chr.17: 82252970-82252970 on Build GRCh38
rs956657158 Chr.17: 82253116-82253116 on Build GRCh38
rs36058840 Chr.17: 82253098-82253099 on Build GRCh38
rs747131812 Chr.17: 82253101-82253101 on Build GRCh38
rs745323845 Chr.17: 82253001-82253001 on Build GRCh38
rs917300671 Chr.17: 82252980-82252980 on Build GRCh38
rs145619785 Chr.17: 82253137-82253137 on Build GRCh38
rs370843744 Chr.17: 82253098-82253098 on Build GRCh38
rs201785865 Chr.17: 82253053-82253053 on Build GRCh38
rs373577958 Chr.17: 82253041-82253041 on Build GRCh38

View Full Product Details