Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49174693-49174964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_011519958.2 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4418779 c.1059A>G p.T353T Chr.11: 49174938-49174938 on Build GRCh38
6133353 c.1066T>G p.Y356D Chr.11: 49174931-49174931 on Build GRCh38
3448218 c.1090G>A p.G364R Chr.11: 49174907-49174907 on Build GRCh38
927687 c.1072G>A p.V358M Chr.11: 49174925-49174925 on Build GRCh38
3448219 c.1056G>A p.V352V Chr.11: 49174941-49174941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772103271 Chr.11: 49174856-49174856 on Build GRCh38
rs751121376 Chr.11: 49174903-49174903 on Build GRCh38
rs775572028 Chr.11: 49174872-49174872 on Build GRCh38
rs140577100 Chr.11: 49174904-49174904 on Build GRCh38
rs1010915861 Chr.11: 49174771-49174771 on Build GRCh38
rs375313419 Chr.11: 49174902-49174902 on Build GRCh38
rs1055466981 Chr.11: 49174739-49174739 on Build GRCh38
rs563801806 Chr.11: 49174885-49174885 on Build GRCh38
rs564957397 Chr.11: 49174807-49174807 on Build GRCh38
rs996418289 Chr.11: 49174817-49174817 on Build GRCh38
rs543920314 Chr.11: 49174882-49174882 on Build GRCh38
rs545141977 Chr.11: 49174784-49174784 on Build GRCh38
rs376770386 Chr.11: 49174919-49174919 on Build GRCh38
rs878865039 Chr.11: 49174770-49174770 on Build GRCh38
rs747804638 Chr.11: 49174909-49174909 on Build GRCh38
rs749660690 Chr.11: 49174841-49174841 on Build GRCh38
rs760831552 Chr.11: 49174880-49174880 on Build GRCh38
rs780980012 Chr.11: 49174906-49174906 on Build GRCh38
rs1019099469 Chr.11: 49174785-49174785 on Build GRCh38
rs1042462986 Chr.11: 49174778-49174781 on Build GRCh38
rs34943739 Chr.11: 49174780-49174781 on Build GRCh38
rs771300656 Chr.11: 49174911-49174911 on Build GRCh38
rs746340486 Chr.11: 49174924-49174924 on Build GRCh38
rs4929893 Chr.11: 49174780-49174780 on Build GRCh38
rs372218227 Chr.11: 49174852-49174852 on Build GRCh38
rs764989225 Chr.11: 49174886-49174886 on Build GRCh38
rs774899532 Chr.11: 49174847-49174847 on Build GRCh38
rs4929894 Chr.11: 49174781-49174781 on Build GRCh38
rs529300272 Chr.11: 49174891-49174891 on Build GRCh38
rs533433837 Chr.11: 49174812-49174812 on Build GRCh38
rs893789129 Chr.11: 49174743-49174743 on Build GRCh38
rs758439576 Chr.11: 49174901-49174901 on Build GRCh38
rs965258203 Chr.11: 49174805-49174805 on Build GRCh38
rs879273667 Chr.11: 49174780-49174781 on Build GRCh38
rs764302751 Chr.11: 49174881-49174881 on Build GRCh38
rs202716 Chr.11: 49174938-49174938 on Build GRCh38
rs771259407 Chr.11: 49174846-49174846 on Build GRCh38

View Full Product Details