Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10767836-10768103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUBP1
Entrez Gene ID
4682
Gene Name
nucleotide binding protein 1
Gene Aliases
NBP, NBP1, NBP35
UniGene
Hs.81469
Gene Chromosome Location
Chr.16: 10743164-10769351 on Build GRCh38
Transcripts
NM_001278506.1 NM_001323594.1 NM_001323595.1 NM_001323596.1 NM_001323597.1 NM_002484.3 XM_017023252.1 NM_001079512.3 NM_001318873.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1 XM_006720944.3 XM_011522654.2 XM_017023649.1 XM_017023650.1 XM_017023651.1

Gene Symbol
TVP23A
Entrez Gene ID
780776
Gene Name
trans-golgi network vesicle protein 23 homolog A
Gene Aliases
FAM18A, YDR084C
UniGene
Hs.371576
Gene Chromosome Location
Chr.16: 10758034-10818902 on Build GRCh38
Transcripts
NM_001278506.1 NM_001323594.1 NM_001323595.1 NM_001323596.1 NM_001323597.1 NM_002484.3 XM_017023252.1 NM_001079512.3 NM_001318873.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1 XM_006720944.3 XM_011522654.2 XM_017023649.1 XM_017023650.1 XM_017023651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6814426 c.845_846insT p.I285fs*2 Chr.16: 10767973-10767974 on Build GRCh38
5722398 c.870C>T p.S290S Chr.16: 10767998-10767998 on Build GRCh38
5713976 c.833A>G p.D278G Chr.16: 10767961-10767961 on Build GRCh38
1640386 c.821-9delT p.? Chr.16: 10767940-10767940 on Build GRCh38
4578649 c.859G>A p.A287T Chr.16: 10767987-10767987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749504011 Chr.16: 10768047-10768047 on Build GRCh38
rs951988118 Chr.16: 10767898-10767898 on Build GRCh38
rs1023985879 Chr.16: 10767858-10767858 on Build GRCh38
rs942501309 Chr.16: 10768020-10768020 on Build GRCh38
rs370645112 Chr.16: 10767927-10767927 on Build GRCh38
rs143719398 Chr.16: 10767982-10767982 on Build GRCh38
rs750371999 Chr.16: 10767931-10767934 on Build GRCh38
rs748371571 Chr.16: 10767953-10767954 on Build GRCh38
rs2233540 Chr.16: 10767859-10767859 on Build GRCh38
rs780064255 Chr.16: 10767949-10767950 on Build GRCh38
rs2302710 Chr.16: 10768066-10768066 on Build GRCh38
rs969427806 Chr.16: 10767872-10767872 on Build GRCh38
rs913127148 Chr.16: 10767943-10767943 on Build GRCh38
rs748308255 Chr.16: 10767964-10767964 on Build GRCh38
rs376176113 Chr.16: 10767922-10767922 on Build GRCh38
rs777750947 Chr.16: 10768045-10768045 on Build GRCh38
rs766747290 Chr.16: 10768007-10768007 on Build GRCh38
rs747491057 Chr.16: 10767956-10767956 on Build GRCh38
rs184643867 Chr.16: 10767894-10767894 on Build GRCh38
rs781740389 Chr.16: 10768031-10768031 on Build GRCh38
rs764677761 Chr.16: 10767988-10767988 on Build GRCh38
rs553999210 Chr.16: 10767870-10767870 on Build GRCh38
rs929615014 Chr.16: 10768058-10768058 on Build GRCh38
rs760645512 Chr.16: 10767916-10767916 on Build GRCh38
rs746737850 Chr.16: 10768065-10768065 on Build GRCh38
rs368639034 Chr.16: 10768006-10768006 on Build GRCh38
rs1055746487 Chr.16: 10768040-10768040 on Build GRCh38
rs756175219 Chr.16: 10768044-10768044 on Build GRCh38
rs148122963 Chr.16: 10767987-10767987 on Build GRCh38
rs1038522348 Chr.16: 10768049-10768049 on Build GRCh38
rs750057791 Chr.16: 10767931-10767931 on Build GRCh38
rs755540705 Chr.16: 10768018-10768018 on Build GRCh38
rs764774759 Chr.16: 10768081-10768081 on Build GRCh38
rs767234408 Chr.16: 10767909-10767909 on Build GRCh38
rs758186337 Chr.16: 10767940-10767940 on Build GRCh38
rs770156675 Chr.16: 10768032-10768035 on Build GRCh38
rs750052496 Chr.16: 10768001-10768001 on Build GRCh38
rs773431831 Chr.16: 10767972-10767972 on Build GRCh38
rs769338852 Chr.16: 10767958-10767958 on Build GRCh38
rs781411477 Chr.16: 10767960-10767960 on Build GRCh38
rs776504881 Chr.16: 10768070-10768070 on Build GRCh38
rs910554548 Chr.16: 10767936-10767936 on Build GRCh38
rs904138379 Chr.16: 10767991-10767991 on Build GRCh38
rs769774616 Chr.16: 10767970-10767970 on Build GRCh38
rs761688745 Chr.16: 10768074-10768074 on Build GRCh38
rs148922151 Chr.16: 10767973-10767973 on Build GRCh38
rs771116449 Chr.16: 10768055-10768055 on Build GRCh38
rs2233541 Chr.16: 10768057-10768057 on Build GRCh38
rs145538631 Chr.16: 10768035-10768035 on Build GRCh38
rs758741535 Chr.16: 10767935-10767935 on Build GRCh38
rs376114818 Chr.16: 10767986-10767986 on Build GRCh38
rs560891578 Chr.16: 10768017-10768017 on Build GRCh38
rs572976431 Chr.16: 10767871-10767871 on Build GRCh38
rs771865092 Chr.16: 10767978-10767978 on Build GRCh38
rs763571135 Chr.16: 10767917-10767917 on Build GRCh38
rs764083699 Chr.16: 10767984-10767984 on Build GRCh38
rs752635308 Chr.16: 10767911-10767911 on Build GRCh38
rs780314321 Chr.16: 10767948-10767948 on Build GRCh38

View Full Product Details