Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68277932-68278205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
-
UniGene
Hs.437249
Gene Chromosome Location
Chr.17: 68259089-68452000 on Build GRCh38
Transcripts
NM_014960.4 XM_005257170.3 XM_005257172.3 XM_006721777.3 XM_006721779.3 XM_011524535.2 XM_011524538.2 XM_011524540.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_011524545.2 XM_017024361.1 XM_017024366.1 NM_001174166.1 NM_004694.4 XM_005257789.3 XM_011525461.2 XM_017025291.1 XM_017025292.1

Gene Symbol
SLC16A6
Entrez Gene ID
9120
Gene Name
solute carrier family 16 member 6
Gene Aliases
MCT6, MCT7
UniGene
Hs.42645
Gene Chromosome Location
Chr.17: 68267026-68291264 on Build GRCh38
Transcripts
NM_014960.4 XM_005257170.3 XM_005257172.3 XM_006721777.3 XM_006721779.3 XM_011524535.2 XM_011524538.2 XM_011524540.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_011524545.2 XM_017024361.1 XM_017024366.1 NM_001174166.1 NM_004694.4 XM_005257789.3 XM_011525461.2 XM_017025291.1 XM_017025292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1729136 c.210T>A p.C70* Chr.17: 68278111-68278111 on Build GRCh38
5624360 c.118G>T p.G40C Chr.17: 68278203-68278203 on Build GRCh38
1225963 c.172G>A p.E58K Chr.17: 68278149-68278149 on Build GRCh38
3388110 c.219C>A p.V73V Chr.17: 68278102-68278102 on Build GRCh38
983395 c.123C>A p.I41I Chr.17: 68278198-68278198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952546800 Chr.17: 68278013-68278013 on Build GRCh38
rs782626604 Chr.17: 68278057-68278058 on Build GRCh38
rs139246850 Chr.17: 68278014-68278014 on Build GRCh38
rs781920926 Chr.17: 68278063-68278063 on Build GRCh38
rs377276849 Chr.17: 68278073-68278073 on Build GRCh38
rs782551003 Chr.17: 68278114-68278114 on Build GRCh38
rs961057412 Chr.17: 68278049-68278049 on Build GRCh38
rs782036328 Chr.17: 68278054-68278054 on Build GRCh38
rs369400870 Chr.17: 68278060-68278060 on Build GRCh38
rs369768355 Chr.17: 68278162-68278162 on Build GRCh38
rs537004480 Chr.17: 68278053-68278053 on Build GRCh38
rs917538465 Chr.17: 68278088-68278088 on Build GRCh38
rs547318076 Chr.17: 68277988-68277988 on Build GRCh38
rs573622082 Chr.17: 68278008-68278008 on Build GRCh38
rs996734299 Chr.17: 68277982-68277982 on Build GRCh38
rs782449635 Chr.17: 68278128-68278128 on Build GRCh38
rs1015277636 Chr.17: 68278045-68278045 on Build GRCh38
rs781878582 Chr.17: 68278102-68278102 on Build GRCh38
rs782776397 Chr.17: 68278112-68278112 on Build GRCh38
rs139353004 Chr.17: 68278157-68278157 on Build GRCh38
rs901086170 Chr.17: 68277981-68277981 on Build GRCh38
rs555069317 Chr.17: 68278025-68278025 on Build GRCh38
rs782735066 Chr.17: 68278074-68278074 on Build GRCh38
rs570078233 Chr.17: 68278031-68278031 on Build GRCh38
rs782259594 Chr.17: 68278052-68278052 on Build GRCh38
rs782153368 Chr.17: 68278059-68278059 on Build GRCh38
rs782516882 Chr.17: 68278183-68278183 on Build GRCh38
rs782604051 Chr.17: 68278153-68278153 on Build GRCh38

View Full Product Details