Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50288139-50288412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY7
Entrez Gene ID
113
Gene Name
adenylate cyclase 7
Gene Aliases
AC7
UniGene
Hs.513578
Gene Chromosome Location
Chr.16: 50246158-50318135 on Build GRCh38
Transcripts
NM_001114.4 NM_001286057.1 XM_011522835.2 XM_011522836.2 XM_011522837.2 XM_011522838.2 XM_011522839.2 XM_011522840.2 XM_011522842.2 XM_017022895.1 XM_017022896.1 XM_017022897.1 XM_017022898.1 XM_017022899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227666 c.17G>A p.R6H Chr.16: 50288196-50288196 on Build GRCh38
6002021 c.142G>A p.A48T Chr.16: 50288321-50288321 on Build GRCh38
7354546 c.151A>G p.I51V Chr.16: 50288330-50288330 on Build GRCh38
7322608 c.143C>A p.A48D Chr.16: 50288322-50288322 on Build GRCh38
3948573 c.30C>T p.N10N Chr.16: 50288209-50288209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201090410 Chr.16: 50288306-50288306 on Build GRCh38
rs757159174 Chr.16: 50288255-50288255 on Build GRCh38
rs895114920 Chr.16: 50288196-50288196 on Build GRCh38
rs758047984 Chr.16: 50288224-50288224 on Build GRCh38
rs77518645 Chr.16: 50288262-50288262 on Build GRCh38
rs764927746 Chr.16: 50288195-50288195 on Build GRCh38
rs759080085 Chr.16: 50288383-50288383 on Build GRCh38
rs982870279 Chr.16: 50288277-50288277 on Build GRCh38
rs764318741 Chr.16: 50288202-50288204 on Build GRCh38
rs1042132967 Chr.16: 50288327-50288327 on Build GRCh38
rs371889230 Chr.16: 50288318-50288318 on Build GRCh38
rs751373586 Chr.16: 50288245-50288245 on Build GRCh38
rs1034181278 Chr.16: 50288354-50288354 on Build GRCh38
rs367883473 Chr.16: 50288209-50288209 on Build GRCh38
rs752356585 Chr.16: 50288216-50288216 on Build GRCh38
rs112244195 Chr.16: 50288359-50288359 on Build GRCh38
rs367714919 Chr.16: 50288317-50288317 on Build GRCh38
rs765710324 Chr.16: 50288365-50288365 on Build GRCh38
rs945972138 Chr.16: 50288363-50288363 on Build GRCh38
rs932235843 Chr.16: 50288313-50288313 on Build GRCh38
rs756799073 Chr.16: 50288348-50288348 on Build GRCh38
rs764582125 Chr.16: 50288384-50288384 on Build GRCh38
rs909610076 Chr.16: 50288278-50288278 on Build GRCh38
rs28514808 Chr.16: 50288289-50288289 on Build GRCh38
rs1000314822 Chr.16: 50288338-50288338 on Build GRCh38
rs1025148390 Chr.16: 50288215-50288215 on Build GRCh38
rs544253864 Chr.16: 50288379-50288379 on Build GRCh38
rs762361791 Chr.16: 50288164-50288164 on Build GRCh38
rs769023727 Chr.16: 50288307-50288307 on Build GRCh38
rs772608421 Chr.16: 50288367-50288367 on Build GRCh38
rs199714954 Chr.16: 50288244-50288244 on Build GRCh38
rs745950378 Chr.16: 50288274-50288274 on Build GRCh38
rs1010212571 Chr.16: 50288323-50288323 on Build GRCh38
rs61731918 Chr.16: 50288258-50288258 on Build GRCh38
rs144769944 Chr.16: 50288305-50288305 on Build GRCh38
rs904592690 Chr.16: 50288336-50288336 on Build GRCh38
rs557064062 Chr.16: 50288163-50288163 on Build GRCh38
rs925748743 Chr.16: 50288378-50288378 on Build GRCh38
rs765896604 Chr.16: 50288239-50288239 on Build GRCh38
rs200299180 Chr.16: 50288238-50288238 on Build GRCh38
rs139493357 Chr.16: 50288179-50288179 on Build GRCh38
rs1028742584 Chr.16: 50288290-50288290 on Build GRCh38
rs774869842 Chr.16: 50288162-50288162 on Build GRCh38
rs867361478 Chr.16: 50288183-50288183 on Build GRCh38
rs369539863 Chr.16: 50288193-50288193 on Build GRCh38
rs537134395 Chr.16: 50288161-50288161 on Build GRCh38

View Full Product Details