Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44995557-44995812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM179B
Entrez Gene ID
23116
Gene Name
family with sequence similarity 179 member B
Gene Aliases
KIAA0423
UniGene
Hs.371078
Gene Chromosome Location
Chr.14: 44962190-45074431 on Build GRCh38
Transcripts
NM_001308120.1 NM_015091.3 NR_131765.1 XM_011536571.1 XM_017021098.1 XM_017021099.1 XM_017021100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382508 c.2074G>A p.A692T Chr.14: 44995773-44995773 on Build GRCh38
1515447 c.2110G>A p.D704N Chr.14: 44995809-44995809 on Build GRCh38
4854736 c.2059G>A p.D687N Chr.14: 44995758-44995758 on Build GRCh38
7203005 c.2075C>T p.A692V Chr.14: 44995774-44995774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746527999 Chr.14: 44995757-44995757 on Build GRCh38
rs758022429 Chr.14: 44995742-44995742 on Build GRCh38
rs973409322 Chr.14: 44995756-44995756 on Build GRCh38
rs1048373149 Chr.14: 44995687-44995687 on Build GRCh38
rs761113576 Chr.14: 44995700-44995700 on Build GRCh38
rs1018344825 Chr.14: 44995778-44995778 on Build GRCh38
rs929883758 Chr.14: 44995669-44995669 on Build GRCh38
rs757588236 Chr.14: 44995723-44995723 on Build GRCh38
rs764572354 Chr.14: 44995712-44995712 on Build GRCh38
rs547200612 Chr.14: 44995592-44995592 on Build GRCh38
rs949487778 Chr.14: 44995622-44995622 on Build GRCh38
rs916695196 Chr.14: 44995611-44995611 on Build GRCh38
rs1046244610 Chr.14: 44995640-44995640 on Build GRCh38
rs754589916 Chr.14: 44995784-44995784 on Build GRCh38
rs370598945 Chr.14: 44995736-44995736 on Build GRCh38
rs754410964 Chr.14: 44995720-44995720 on Build GRCh38
rs756837446 Chr.14: 44995591-44995591 on Build GRCh38
rs767872811 Chr.14: 44995651-44995651 on Build GRCh38
rs781268924 Chr.14: 44995786-44995786 on Build GRCh38
rs77644097 Chr.14: 44995662-44995662 on Build GRCh38
rs141945491 Chr.14: 44995754-44995754 on Build GRCh38

View Full Product Details