Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67441951-67442176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1B
Entrez Gene ID
57175
Gene Name
coronin 1B
Gene Aliases
CORONIN-2
UniGene
Hs.6191
Gene Chromosome Location
Chr.11: 67438047-67443821 on Build GRCh38
Transcripts
NM_001018070.2 NM_020441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634206 c.295G>T p.A99S Chr.11: 67441995-67441995 on Build GRCh38
3687790 c.231G>A p.T77T Chr.11: 67442059-67442059 on Build GRCh38
6634205 c.295G>T p.A99S Chr.11: 67441995-67441995 on Build GRCh38
7077168 c.228G>A p.P76P Chr.11: 67442062-67442062 on Build GRCh38
4786743 c.286G>A p.E96K Chr.11: 67442004-67442004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038855963 Chr.11: 67442086-67442086 on Build GRCh38
rs780259406 Chr.11: 67441986-67441988 on Build GRCh38
rs781711099 Chr.11: 67442082-67442082 on Build GRCh38
rs764414306 Chr.11: 67442130-67442130 on Build GRCh38
rs796147666 Chr.11: 67442034-67442034 on Build GRCh38
rs369704819 Chr.11: 67442083-67442083 on Build GRCh38
rs774302969 Chr.11: 67442008-67442008 on Build GRCh38
rs201571103 Chr.11: 67441998-67441998 on Build GRCh38
rs758707123 Chr.11: 67441973-67441973 on Build GRCh38
rs143715095 Chr.11: 67442005-67442005 on Build GRCh38
rs772122884 Chr.11: 67442107-67442107 on Build GRCh38
rs111451405 Chr.11: 67442044-67442044 on Build GRCh38
rs761635879 Chr.11: 67442014-67442014 on Build GRCh38
rs752869875 Chr.11: 67442058-67442058 on Build GRCh38
rs748043438 Chr.11: 67442106-67442106 on Build GRCh38
rs942935140 Chr.11: 67441985-67441985 on Build GRCh38
rs200415950 Chr.11: 67441989-67441989 on Build GRCh38
rs1041737087 Chr.11: 67442157-67442157 on Build GRCh38
rs773299865 Chr.11: 67442110-67442110 on Build GRCh38
rs753102227 Chr.11: 67441972-67441972 on Build GRCh38
rs757671047 Chr.11: 67442081-67442081 on Build GRCh38
rs1024369675 Chr.11: 67442154-67442154 on Build GRCh38
rs765501761 Chr.11: 67442045-67442045 on Build GRCh38
rs539121732 Chr.11: 67442144-67442144 on Build GRCh38
rs975987715 Chr.11: 67442132-67442132 on Build GRCh38
rs567987315 Chr.11: 67442059-67442059 on Build GRCh38
rs200915118 Chr.11: 67442125-67442125 on Build GRCh38
rs749918070 Chr.11: 67442084-67442084 on Build GRCh38
rs768338805 Chr.11: 67442007-67442007 on Build GRCh38
rs754173098 Chr.11: 67441987-67441988 on Build GRCh38
rs535013161 Chr.11: 67442060-67442060 on Build GRCh38
rs565558736 Chr.11: 67442098-67442098 on Build GRCh38
rs752039855 Chr.11: 67441984-67441984 on Build GRCh38
rs768446687 Chr.11: 67442099-67442099 on Build GRCh38
rs778542733 Chr.11: 67442105-67442105 on Build GRCh38
rs778279504 Chr.11: 67441977-67441977 on Build GRCh38
rs745881144 Chr.11: 67442116-67442116 on Build GRCh38
rs114276194 Chr.11: 67442156-67442156 on Build GRCh38
rs199638846 Chr.11: 67442078-67442078 on Build GRCh38
rs142190430 Chr.11: 67442087-67442087 on Build GRCh38
rs377469268 Chr.11: 67442129-67442129 on Build GRCh38
rs920254806 Chr.11: 67442029-67442029 on Build GRCh38
rs770027209 Chr.11: 67442124-67442124 on Build GRCh38
rs772178115 Chr.11: 67442025-67442025 on Build GRCh38
rs779218228 Chr.11: 67442106-67442107 on Build GRCh38
rs971518352 Chr.11: 67442155-67442155 on Build GRCh38
rs769428845 Chr.11: 67442004-67442004 on Build GRCh38
rs780654908 Chr.11: 67441990-67441990 on Build GRCh38
rs753065408 Chr.11: 67442069-67442070 on Build GRCh38
rs752001928 Chr.11: 67442062-67442062 on Build GRCh38
rs748057679 Chr.11: 67442134-67442134 on Build GRCh38

View Full Product Details