Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41116332-41116569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2F1
Entrez Gene ID
1572
Gene Name
cytochrome P450 family 2 subfamily F member 1
Gene Aliases
C2F1, CYP2F, CYPIIF1
UniGene
Hs.558318
Gene Chromosome Location
Chr.19: 41114292-41128381 on Build GRCh38
Transcripts
NM_000774.4 NR_135528.1 XM_011526551.2 XM_011526552.2 XM_011526553.2 XM_011526554.2 XM_011526555.2 XM_017026384.1 XM_017026385.1 XM_017026386.1 XM_017026387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
996925 c.154C>A p.L52M Chr.19: 41116342-41116342 on Build GRCh38
6084927 c.173T>A p.L58Q Chr.19: 41116456-41116456 on Build GRCh38
4935280 c.228G>A p.V76V Chr.19: 41116511-41116511 on Build GRCh38
5560520 c.153G>T p.M51I Chr.19: 41116341-41116341 on Build GRCh38
4078415 c.151A>C p.M51L Chr.19: 41116339-41116339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772249109 Chr.19: 41116458-41116458 on Build GRCh38
rs779989368 Chr.19: 41116460-41116460 on Build GRCh38
rs766915556 Chr.19: 41116413-41116413 on Build GRCh38
rs377017877 Chr.19: 41116389-41116389 on Build GRCh38
rs757071675 Chr.19: 41116433-41116433 on Build GRCh38
rs745767766 Chr.19: 41116450-41116450 on Build GRCh38
rs747044071 Chr.19: 41116464-41116464 on Build GRCh38
rs762195176 Chr.19: 41116503-41116503 on Build GRCh38
rs200901140 Chr.19: 41116390-41116390 on Build GRCh38
rs989878161 Chr.19: 41116434-41116434 on Build GRCh38
rs776783483 Chr.19: 41116487-41116487 on Build GRCh38
rs776980284 Chr.19: 41116380-41116380 on Build GRCh38
rs915311064 Chr.19: 41116437-41116437 on Build GRCh38
rs760208996 Chr.19: 41116530-41116530 on Build GRCh38
rs759127483 Chr.19: 41116398-41116398 on Build GRCh38
rs755721234 Chr.19: 41116418-41116418 on Build GRCh38
rs977519814 Chr.19: 41116475-41116475 on Build GRCh38
rs753703363 Chr.19: 41116430-41116430 on Build GRCh38
rs777504485 Chr.19: 41116420-41116420 on Build GRCh38
rs928536385 Chr.19: 41116501-41116501 on Build GRCh38
rs144909127 Chr.19: 41116506-41116506 on Build GRCh38
rs777323567 Chr.19: 41116521-41116521 on Build GRCh38
rs762105225 Chr.19: 41116383-41116383 on Build GRCh38
rs763356097 Chr.19: 41116520-41116520 on Build GRCh38
rs373894342 Chr.19: 41116379-41116379 on Build GRCh38
rs1039303112 Chr.19: 41116547-41116547 on Build GRCh38
rs937837637 Chr.19: 41116369-41116369 on Build GRCh38
rs74824096 Chr.19: 41116362-41116362 on Build GRCh38
rs145103345 Chr.19: 41116504-41116504 on Build GRCh38
rs902123416 Chr.19: 41116376-41116376 on Build GRCh38
rs772011895 Chr.19: 41116364-41116364 on Build GRCh38
rs775829771 Chr.19: 41116368-41116368 on Build GRCh38
rs757994193 Chr.19: 41116465-41116466 on Build GRCh38
rs768879115 Chr.19: 41116484-41116484 on Build GRCh38
rs779032097 Chr.19: 41116449-41116449 on Build GRCh38
rs752465443 Chr.19: 41116429-41116429 on Build GRCh38

View Full Product Details