Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177515049-177515286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX41
Entrez Gene ID
51428
Gene Name
DEAD-box helicase 41
Gene Aliases
ABS, MPLPF
UniGene
Hs.484288
Gene Chromosome Location
Chr.5: 177511577-177517326 on Build GRCh38
Transcripts
NM_001321732.1 NM_001321830.1 NM_016222.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7208614 c.601A>T p.I201F Chr.5: 177515229-177515229 on Build GRCh38
7203436 c.590A>G p.K197R Chr.5: 177515240-177515240 on Build GRCh38
4155779 c.591G>A p.K197K Chr.5: 177515239-177515239 on Build GRCh38
6641820 c.660C>T p.D220D Chr.5: 177515054-177515054 on Build GRCh38
1486637 c.637C>G p.P213A Chr.5: 177515193-177515193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541153895 Chr.5: 177515197-177515197 on Build GRCh38
rs781436670 Chr.5: 177515203-177515203 on Build GRCh38
rs145085939 Chr.5: 177515233-177515233 on Build GRCh38
rs755198680 Chr.5: 177515088-177515088 on Build GRCh38
rs749253420 Chr.5: 177515111-177515111 on Build GRCh38
rs149122003 Chr.5: 177515239-177515239 on Build GRCh38
rs778763328 Chr.5: 177515245-177515245 on Build GRCh38
rs371633755 Chr.5: 177515093-177515093 on Build GRCh38
rs200056121 Chr.5: 177515162-177515162 on Build GRCh38
rs780120166 Chr.5: 177515079-177515079 on Build GRCh38
rs905341773 Chr.5: 177515222-177515222 on Build GRCh38
rs775024380 Chr.5: 177515166-177515166 on Build GRCh38
rs935314360 Chr.5: 177515118-177515118 on Build GRCh38
rs773904073 Chr.5: 177515119-177515119 on Build GRCh38
rs759005362 Chr.5: 177515135-177515135 on Build GRCh38
rs771510721 Chr.5: 177515247-177515247 on Build GRCh38
rs781318967 Chr.5: 177515089-177515089 on Build GRCh38
rs962423100 Chr.5: 177515191-177515191 on Build GRCh38
rs1013249905 Chr.5: 177515087-177515087 on Build GRCh38
rs920927202 Chr.5: 177515244-177515244 on Build GRCh38
rs1004241531 Chr.5: 177515253-177515253 on Build GRCh38
rs773536438 Chr.5: 177515263-177515263 on Build GRCh38
rs369566052 Chr.5: 177515186-177515186 on Build GRCh38
rs376574336 Chr.5: 177515115-177515115 on Build GRCh38
rs34567318 Chr.5: 177515203-177515204 on Build GRCh38
rs755036127 Chr.5: 177515200-177515200 on Build GRCh38
rs747126487 Chr.5: 177515080-177515080 on Build GRCh38
rs775458261 Chr.5: 177515113-177515115 on Build GRCh38
rs375274366 Chr.5: 177515254-177515254 on Build GRCh38
rs114125242 Chr.5: 177515090-177515090 on Build GRCh38
rs773221580 Chr.5: 177515098-177515098 on Build GRCh38
rs982554644 Chr.5: 177515137-177515137 on Build GRCh38
rs760334582 Chr.5: 177515261-177515261 on Build GRCh38
rs755920162 Chr.5: 177515231-177515231 on Build GRCh38
rs373575560 Chr.5: 177515214-177515214 on Build GRCh38
rs745329544 Chr.5: 177515246-177515246 on Build GRCh38
rs769733379 Chr.5: 177515107-177515116 on Build GRCh38
rs768353496 Chr.5: 177515262-177515262 on Build GRCh38
rs759325258 Chr.5: 177515236-177515236 on Build GRCh38
rs766913824 Chr.5: 177515192-177515192 on Build GRCh38
rs749438678 Chr.5: 177515177-177515178 on Build GRCh38
rs762124039 Chr.5: 177515176-177515176 on Build GRCh38
rs573545872 Chr.5: 177515187-177515187 on Build GRCh38

View Full Product Details