Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156294544-156294789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMP
Entrez Gene ID
112770
Gene Name
glycosylated lysosomal membrane protein
Gene Aliases
C1orf85, NCU-G1
UniGene
Hs.202522
Gene Chromosome Location
Chr.1: 156292687-156295689 on Build GRCh38
Transcripts
NM_001256604.1 NM_001256605.1 NM_001256608.1 NM_001256609.1 NM_144580.2 XM_011509117.2 XM_017000149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1983305 c.365T>C p.L122P Chr.1: 156294772-156294772 on Build GRCh38
5818051 c.385G>A p.E129K Chr.1: 156294559-156294559 on Build GRCh38
5776999 c.377G>A p.R126K Chr.1: 156294760-156294760 on Build GRCh38
5818052 c.127G>A p.E43K Chr.1: 156294559-156294559 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs989460748 Chr.1: 156294617-156294617 on Build GRCh38
rs769327921 Chr.1: 156294678-156294678 on Build GRCh38
rs939535977 Chr.1: 156294700-156294700 on Build GRCh38
rs765387547 Chr.1: 156294708-156294708 on Build GRCh38
rs766213115 Chr.1: 156294581-156294581 on Build GRCh38
rs768179765 Chr.1: 156294666-156294666 on Build GRCh38
rs957270292 Chr.1: 156294599-156294599 on Build GRCh38
rs780426971 Chr.1: 156294658-156294658 on Build GRCh38
rs559674341 Chr.1: 156294694-156294694 on Build GRCh38
rs71591935 Chr.1: 156294581-156294582 on Build GRCh38
rs747471461 Chr.1: 156294665-156294665 on Build GRCh38
rs774450784 Chr.1: 156294686-156294686 on Build GRCh38
rs545200384 Chr.1: 156294710-156294710 on Build GRCh38
rs780660881 Chr.1: 156294675-156294675 on Build GRCh38
rs1022311433 Chr.1: 156294643-156294643 on Build GRCh38
rs114160426 Chr.1: 156294698-156294698 on Build GRCh38
rs150363388 Chr.1: 156294654-156294654 on Build GRCh38
rs1011010746 Chr.1: 156294737-156294737 on Build GRCh38
rs771534839 Chr.1: 156294729-156294730 on Build GRCh38
rs554917532 Chr.1: 156294660-156294660 on Build GRCh38
rs756923387 Chr.1: 156294640-156294640 on Build GRCh38
rs764219490 Chr.1: 156294589-156294589 on Build GRCh38
rs760842853 Chr.1: 156294586-156294586 on Build GRCh38
rs745426584 Chr.1: 156294725-156294726 on Build GRCh38
rs750589221 Chr.1: 156294709-156294709 on Build GRCh38
rs762671649 Chr.1: 156294680-156294680 on Build GRCh38
rs565710258 Chr.1: 156294615-156294615 on Build GRCh38
rs990974614 Chr.1: 156294759-156294759 on Build GRCh38
rs573155616 Chr.1: 156294679-156294679 on Build GRCh38
rs779400581 Chr.1: 156294638-156294638 on Build GRCh38
rs753530342 Chr.1: 156294639-156294640 on Build GRCh38
rs557162682 Chr.1: 156294580-156294580 on Build GRCh38
rs757521230 Chr.1: 156294635-156294635 on Build GRCh38
rs779992047 Chr.1: 156294743-156294750 on Build GRCh38
rs764309401 Chr.1: 156294692-156294692 on Build GRCh38
rs752112358 Chr.1: 156294758-156294758 on Build GRCh38
rs541284459 Chr.1: 156294691-156294691 on Build GRCh38
rs758727289 Chr.1: 156294657-156294657 on Build GRCh38
rs1040358262 Chr.1: 156294683-156294683 on Build GRCh38
rs778519824 Chr.1: 156294674-156294675 on Build GRCh38
rs772855023 Chr.1: 156294577-156294577 on Build GRCh38
rs769630354 Chr.1: 156294567-156294567 on Build GRCh38
rs751457868 Chr.1: 156294582-156294582 on Build GRCh38
rs71630623 Chr.1: 156294627-156294627 on Build GRCh38
rs575680686 Chr.1: 156294614-156294614 on Build GRCh38
rs563371300 Chr.1: 156294711-156294711 on Build GRCh38
rs368809546 Chr.1: 156294575-156294575 on Build GRCh38
rs967081145 Chr.1: 156294684-156294684 on Build GRCh38

View Full Product Details