Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151076924-151077171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTK
Entrez Gene ID
10922
Gene Name
Fas activated serine/threonine kinase
Gene Aliases
FAST
UniGene
Hs.647094
Gene Chromosome Location
Chr.7: 151076621-151080883 on Build GRCh38
Transcripts
NM_001258461.1 NM_006712.4 NM_033015.3 XM_005249932.1 XM_005249933.3 XM_011515761.2 XM_011515762.2 XM_011515763.1 XM_017011704.1 XM_017011705.1 XM_017011706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6570300 c.1390G>T p.A464S Chr.7: 151077138-151077138 on Build GRCh38
1488373 c.1491G>C p.S497S Chr.7: 151076964-151076964 on Build GRCh38
7245666 c.1475G>A p.R492Q Chr.7: 151076980-151076980 on Build GRCh38
4161963 c.1458T>G p.H486Q Chr.7: 151076997-151076997 on Build GRCh38
7241935 c.1491G>A p.S497S Chr.7: 151076964-151076964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749421055 Chr.7: 151077046-151077046 on Build GRCh38
rs771377198 Chr.7: 151077078-151077078 on Build GRCh38
rs747990923 Chr.7: 151076953-151076953 on Build GRCh38
rs371380337 Chr.7: 151076990-151076990 on Build GRCh38
rs201888609 Chr.7: 151077143-151077143 on Build GRCh38
rs532948471 Chr.7: 151077083-151077083 on Build GRCh38
rs754383437 Chr.7: 151076943-151076943 on Build GRCh38
rs535070707 Chr.7: 151077079-151077079 on Build GRCh38
rs748390462 Chr.7: 151077138-151077138 on Build GRCh38
rs924056895 Chr.7: 151077113-151077113 on Build GRCh38
rs753396971 Chr.7: 151077010-151077010 on Build GRCh38
rs762393572 Chr.7: 151077100-151077100 on Build GRCh38
rs758330052 Chr.7: 151077036-151077036 on Build GRCh38
rs768577800 Chr.7: 151076985-151076985 on Build GRCh38
rs140806409 Chr.7: 151077136-151077136 on Build GRCh38
rs779249636 Chr.7: 151077131-151077131 on Build GRCh38
rs906204673 Chr.7: 151076981-151076981 on Build GRCh38
rs986078441 Chr.7: 151077052-151077052 on Build GRCh38
rs750271006 Chr.7: 151077126-151077126 on Build GRCh38
rs774003292 Chr.7: 151077044-151077044 on Build GRCh38
rs941805088 Chr.7: 151077037-151077037 on Build GRCh38
rs759082855 Chr.7: 151077080-151077080 on Build GRCh38
rs200153094 Chr.7: 151076984-151076984 on Build GRCh38
rs367561144 Chr.7: 151076965-151076965 on Build GRCh38
rs35223428 Chr.7: 151077003-151077003 on Build GRCh38
rs868369726 Chr.7: 151077093-151077093 on Build GRCh38
rs752137739 Chr.7: 151077087-151077087 on Build GRCh38
rs201230993 Chr.7: 151077034-151077034 on Build GRCh38
rs369209406 Chr.7: 151077068-151077068 on Build GRCh38
rs761176338 Chr.7: 151077005-151077005 on Build GRCh38
rs769991300 Chr.7: 151076980-151076980 on Build GRCh38
rs200625890 Chr.7: 151077107-151077107 on Build GRCh38
rs751459242 Chr.7: 151077112-151077112 on Build GRCh38
rs201972478 Chr.7: 151077033-151077033 on Build GRCh38
rs139030865 Chr.7: 151077061-151077061 on Build GRCh38
rs756979216 Chr.7: 151077043-151077043 on Build GRCh38
rs534704710 Chr.7: 151077149-151077149 on Build GRCh38
rs199875725 Chr.7: 151077110-151077110 on Build GRCh38
rs1053984090 Chr.7: 151077122-151077122 on Build GRCh38
rs761264423 Chr.7: 151077069-151077069 on Build GRCh38
rs768176226 Chr.7: 151076994-151076994 on Build GRCh38
rs371534765 Chr.7: 151077124-151077124 on Build GRCh38
rs768971500 Chr.7: 151077060-151077060 on Build GRCh38
rs1130560 Chr.7: 151077103-151077103 on Build GRCh38
rs778520792 Chr.7: 151076948-151076948 on Build GRCh38
rs758413763 Chr.7: 151076964-151076964 on Build GRCh38
rs766859417 Chr.7: 151077008-151077008 on Build GRCh38
rs755148604 Chr.7: 151077127-151077127 on Build GRCh38
rs777747992 Chr.7: 151077039-151077039 on Build GRCh38
rs765009890 Chr.7: 151077084-151077084 on Build GRCh38
rs150140464 Chr.7: 151077148-151077148 on Build GRCh38
rs757207922 Chr.7: 151077116-151077116 on Build GRCh38
rs772689989 Chr.7: 151077067-151077067 on Build GRCh38
rs774510924 Chr.7: 151076989-151076989 on Build GRCh38
rs754684863 Chr.7: 151077030-151077030 on Build GRCh38
rs746874695 Chr.7: 151076967-151076967 on Build GRCh38

View Full Product Details