Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47554641-47554874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_011514449.2 XM_017010641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623659 c.494T>C p.L165S Chr.6: 47554719-47554719 on Build GRCh38
1079997 c.535G>A p.D179N Chr.6: 47554760-47554760 on Build GRCh38
6040728 c.502_503insAC p.T168fs*31 Chr.6: 47554727-47554728 on Build GRCh38
6038949 c.498G>A p.E166E Chr.6: 47554723-47554723 on Build GRCh38
4966994 c.421-1G>T p.? Chr.6: 47554645-47554645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534348943 Chr.6: 47554837-47554837 on Build GRCh38
rs777620356 Chr.6: 47554664-47554664 on Build GRCh38
rs570511925 Chr.6: 47554677-47554677 on Build GRCh38
rs371806727 Chr.6: 47554812-47554812 on Build GRCh38
rs200023127 Chr.6: 47554811-47554811 on Build GRCh38
rs765668592 Chr.6: 47554747-47554747 on Build GRCh38
rs886061520 Chr.6: 47554755-47554755 on Build GRCh38
rs762020246 Chr.6: 47554746-47554746 on Build GRCh38
rs772227398 Chr.6: 47554696-47554696 on Build GRCh38
rs760969524 Chr.6: 47554764-47554764 on Build GRCh38
rs912631923 Chr.6: 47554840-47554840 on Build GRCh38
rs763594204 Chr.6: 47554793-47554793 on Build GRCh38
rs776930343 Chr.6: 47554731-47554731 on Build GRCh38
rs775759148 Chr.6: 47554697-47554697 on Build GRCh38
rs777528378 Chr.6: 47554686-47554691 on Build GRCh38
rs1012340609 Chr.6: 47554759-47554759 on Build GRCh38
rs750946190 Chr.6: 47554692-47554692 on Build GRCh38
rs780272900 Chr.6: 47554818-47554818 on Build GRCh38
rs550136465 Chr.6: 47554800-47554800 on Build GRCh38
rs763501713 Chr.6: 47554776-47554776 on Build GRCh38
rs746499088 Chr.6: 47554785-47554786 on Build GRCh38
rs200598584 Chr.6: 47554785-47554785 on Build GRCh38
rs145518596 Chr.6: 47554672-47554672 on Build GRCh38
rs769062684 Chr.6: 47554727-47554727 on Build GRCh38
rs201727632 Chr.6: 47554815-47554815 on Build GRCh38
rs755750535 Chr.6: 47554792-47554792 on Build GRCh38
rs766983546 Chr.6: 47554780-47554780 on Build GRCh38
rs756960248 Chr.6: 47554810-47554810 on Build GRCh38
rs745830059 Chr.6: 47554680-47554680 on Build GRCh38
rs1004659594 Chr.6: 47554791-47554791 on Build GRCh38
rs754200490 Chr.6: 47554790-47554790 on Build GRCh38
rs757088308 Chr.6: 47554673-47554673 on Build GRCh38
rs747200381 Chr.6: 47554699-47554699 on Build GRCh38
rs566853138 Chr.6: 47554834-47554834 on Build GRCh38
rs555004813 Chr.6: 47554821-47554821 on Build GRCh38
rs758359789 Chr.6: 47554824-47554824 on Build GRCh38

View Full Product Details