Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119112541-119112784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD2L
Entrez Gene ID
9854
Gene Name
C2CD2 like
Gene Aliases
DLNB23, TMEM24
UniGene
Hs.26899
Gene Chromosome Location
Chr.11: 119107061-119118543 on Build GRCh38
Transcripts
NM_001290474.1 NM_014807.4 XM_006718948.3 XM_011543076.2 XM_017018625.1 XM_017018626.1 XM_017018627.1 XM_017018628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172836 c.1220A>G p.Y407C Chr.11: 119112707-119112707 on Build GRCh38
4018471 c.1270C>T p.R424C Chr.11: 119112757-119112757 on Build GRCh38
923754 c.1243C>A p.L415M Chr.11: 119112730-119112730 on Build GRCh38
5714113 c.1212G>A p.E404E Chr.11: 119112609-119112609 on Build GRCh38
3998256 c.1237C>T p.R413W Chr.11: 119112724-119112724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746415496 Chr.11: 119112697-119112697 on Build GRCh38
rs747755293 Chr.11: 119112666-119112666 on Build GRCh38
rs773933044 Chr.11: 119112754-119112754 on Build GRCh38
rs748791627 Chr.11: 119112663-119112663 on Build GRCh38
rs748815354 Chr.11: 119112684-119112684 on Build GRCh38
rs754978548 Chr.11: 119112762-119112762 on Build GRCh38
rs753228335 Chr.11: 119112693-119112693 on Build GRCh38
rs1009603575 Chr.11: 119112620-119112620 on Build GRCh38
rs751306158 Chr.11: 119112650-119112650 on Build GRCh38
rs143682926 Chr.11: 119112606-119112606 on Build GRCh38
rs371031696 Chr.11: 119112667-119112667 on Build GRCh38
rs771581644 Chr.11: 119112584-119112584 on Build GRCh38
rs759327089 Chr.11: 119112755-119112755 on Build GRCh38
rs868123536 Chr.11: 119112589-119112589 on Build GRCh38
rs772557061 Chr.11: 119112709-119112709 on Build GRCh38
rs767956921 Chr.11: 119112653-119112653 on Build GRCh38
rs759996756 Chr.11: 119112600-119112600 on Build GRCh38
rs977928719 Chr.11: 119112752-119112752 on Build GRCh38
rs140734829 Chr.11: 119112641-119112641 on Build GRCh38
rs549880903 Chr.11: 119112646-119112647 on Build GRCh38
rs752828682 Chr.11: 119112612-119112612 on Build GRCh38
rs760748375 Chr.11: 119112716-119112716 on Build GRCh38
rs373042322 Chr.11: 119112725-119112725 on Build GRCh38
rs2239896 Chr.11: 119112724-119112724 on Build GRCh38
rs747885394 Chr.11: 119112560-119112560 on Build GRCh38
rs565924451 Chr.11: 119112623-119112623 on Build GRCh38
rs764066439 Chr.11: 119112631-119112631 on Build GRCh38
rs754746691 Chr.11: 119112659-119112659 on Build GRCh38
rs373719904 Chr.11: 119112634-119112634 on Build GRCh38
rs771512046 Chr.11: 119112685-119112685 on Build GRCh38
rs774972096 Chr.11: 119112696-119112696 on Build GRCh38
rs890713799 Chr.11: 119112573-119112573 on Build GRCh38
rs369410743 Chr.11: 119112592-119112592 on Build GRCh38
rs774952377 Chr.11: 119112758-119112758 on Build GRCh38
rs202229214 Chr.11: 119112757-119112757 on Build GRCh38
rs373100062 Chr.11: 119112568-119112568 on Build GRCh38
rs766335218 Chr.11: 119112649-119112649 on Build GRCh38
rs775979236 Chr.11: 119112711-119112711 on Build GRCh38
rs199820148 Chr.11: 119112569-119112569 on Build GRCh38
rs777093266 Chr.11: 119112672-119112672 on Build GRCh38
rs763360089 Chr.11: 119112571-119112571 on Build GRCh38

View Full Product Details