Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75235406-75235655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMTD1
Entrez Gene ID
118881
Gene Name
catechol-O-methyltransferase domain containing 1
Gene Aliases
-
UniGene
Hs.355333
Gene Chromosome Location
Chr.10: 75233971-75236012 on Build GRCh38
Transcripts
NM_144589.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372832 c.191G>A p.R64Q Chr.10: 75235647-75235647 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369759982 Chr.10: 75235600-75235600 on Build GRCh38
rs759097625 Chr.10: 75235568-75235568 on Build GRCh38
rs552138426 Chr.10: 75235465-75235465 on Build GRCh38
rs397846930 Chr.10: 75235443-75235443 on Build GRCh38
rs770644866 Chr.10: 75235565-75235565 on Build GRCh38
rs41280438 Chr.10: 75235496-75235496 on Build GRCh38
rs749683038 Chr.10: 75235639-75235639 on Build GRCh38
rs771751152 Chr.10: 75235570-75235570 on Build GRCh38
rs763739066 Chr.10: 75235588-75235588 on Build GRCh38
rs527628753 Chr.10: 75235449-75235449 on Build GRCh38
rs767641579 Chr.10: 75235629-75235629 on Build GRCh38
rs753144651 Chr.10: 75235634-75235634 on Build GRCh38
rs556159586 Chr.10: 75235521-75235530 on Build GRCh38
rs192698632 Chr.10: 75235598-75235598 on Build GRCh38
rs373496799 Chr.10: 75235604-75235604 on Build GRCh38
rs375559155 Chr.10: 75235592-75235592 on Build GRCh38
rs957882272 Chr.10: 75235543-75235543 on Build GRCh38
rs1000546971 Chr.10: 75235525-75235525 on Build GRCh38
rs893216502 Chr.10: 75235514-75235514 on Build GRCh38
rs760756869 Chr.10: 75235579-75235579 on Build GRCh38
rs777955867 Chr.10: 75235638-75235638 on Build GRCh38
rs549869111 Chr.10: 75235530-75235530 on Build GRCh38
rs531173148 Chr.10: 75235528-75235528 on Build GRCh38
rs1032195334 Chr.10: 75235508-75235508 on Build GRCh38
rs374644137 Chr.10: 75235558-75235558 on Build GRCh38
rs1040577865 Chr.10: 75235446-75235446 on Build GRCh38
rs943629116 Chr.10: 75235440-75235440 on Build GRCh38
rs568188497 Chr.10: 75235538-75235538 on Build GRCh38
rs775567294 Chr.10: 75235574-75235574 on Build GRCh38
rs889251503 Chr.10: 75235430-75235430 on Build GRCh38
rs776491395 Chr.10: 75235591-75235591 on Build GRCh38
rs773682868 Chr.10: 75235566-75235566 on Build GRCh38
rs997707666 Chr.10: 75235476-75235476 on Build GRCh38
rs760664849 Chr.10: 75235548-75235548 on Build GRCh38

View Full Product Details