Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118756151-118756424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX6
Entrez Gene ID
1656
Gene Name
DEAD-box helicase 6
Gene Aliases
HLR2, P54, RCK
UniGene
Hs.408461
Gene Chromosome Location
Chr.11: 118747763-118791263 on Build GRCh38
Transcripts
NM_001257191.2 NM_004397.5 XM_005271417.3 XM_011542644.1 XM_011542645.1 XM_017017251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6014692 c.1111-4C>T p.? Chr.11: 118756327-118756327 on Build GRCh38
298635 c.1085G>A p.R362Q Chr.11: 118756316-118756316 on Build GRCh38
6642039 c.1142G>A p.R381Q Chr.11: 118756292-118756292 on Build GRCh38
6642037 c.1141C>T p.R381* Chr.11: 118756293-118756293 on Build GRCh38
1177110 c.1118G>A p.R373Q Chr.11: 118756316-118756316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782729363 Chr.11: 118756359-118756359 on Build GRCh38
rs782089328 Chr.11: 118756339-118756339 on Build GRCh38
rs923635270 Chr.11: 118756387-118756387 on Build GRCh38
rs782371749 Chr.11: 118756274-118756274 on Build GRCh38
rs200427531 Chr.11: 118756364-118756364 on Build GRCh38
rs576060897 Chr.11: 118756217-118756217 on Build GRCh38
rs974885233 Chr.11: 118756291-118756291 on Build GRCh38
rs782274778 Chr.11: 118756278-118756278 on Build GRCh38
rs557694300 Chr.11: 118756188-118756188 on Build GRCh38
rs533168978 Chr.11: 118756213-118756213 on Build GRCh38
rs782816895 Chr.11: 118756371-118756371 on Build GRCh38
rs782177348 Chr.11: 118756186-118756186 on Build GRCh38
rs782255367 Chr.11: 118756236-118756236 on Build GRCh38
rs782559020 Chr.11: 118756211-118756211 on Build GRCh38
rs782031324 Chr.11: 118756276-118756276 on Build GRCh38
rs782196571 Chr.11: 118756215-118756215 on Build GRCh38
rs781915724 Chr.11: 118756338-118756338 on Build GRCh38
rs782354421 Chr.11: 118756330-118756330 on Build GRCh38
rs557704768 Chr.11: 118756336-118756336 on Build GRCh38
rs782626975 Chr.11: 118756218-118756218 on Build GRCh38
rs782115935 Chr.11: 118756368-118756368 on Build GRCh38
rs979127660 Chr.11: 118756374-118756374 on Build GRCh38
rs782670170 Chr.11: 118756212-118756212 on Build GRCh38

View Full Product Details