Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30835232-30835493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD5
Entrez Gene ID
79915
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
C17orf41, ELG1, FRAG1
UniGene
Hs.528902
Gene Chromosome Location
Chr.17: 30830932-30896424 on Build GRCh38
Transcripts
NM_024857.4 XM_011525269.2 XM_011525270.2 XM_011525273.2 XM_017025111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5967611 c.1352G>A p.G451D Chr.17: 30835433-30835433 on Build GRCh38
3179366 c.1372A>C p.N458H Chr.17: 30835453-30835453 on Build GRCh38
6606428 c.1252G>T p.G418* Chr.17: 30835333-30835333 on Build GRCh38
7219538 c.1181A>C p.K394T Chr.17: 30835262-30835262 on Build GRCh38
1183865 c.1274A>C p.K425T Chr.17: 30835355-30835355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149829392 Chr.17: 30835331-30835331 on Build GRCh38
rs530437486 Chr.17: 30835299-30835299 on Build GRCh38
rs202223610 Chr.17: 30835352-30835352 on Build GRCh38
rs755109692 Chr.17: 30835333-30835333 on Build GRCh38
rs778334462 Chr.17: 30835390-30835390 on Build GRCh38
rs999234475 Chr.17: 30835366-30835366 on Build GRCh38
rs760447210 Chr.17: 30835320-30835320 on Build GRCh38
rs771428142 Chr.17: 30835384-30835384 on Build GRCh38
rs752691843 Chr.17: 30835336-30835336 on Build GRCh38
rs759042319 Chr.17: 30835345-30835345 on Build GRCh38
rs916281942 Chr.17: 30835383-30835383 on Build GRCh38
rs750280559 Chr.17: 30835309-30835309 on Build GRCh38
rs748989862 Chr.17: 30835400-30835400 on Build GRCh38
rs761858157 Chr.17: 30835297-30835297 on Build GRCh38
rs768242074 Chr.17: 30835265-30835265 on Build GRCh38
rs773951046 Chr.17: 30835458-30835458 on Build GRCh38
rs147926164 Chr.17: 30835351-30835351 on Build GRCh38
rs770018311 Chr.17: 30835399-30835401 on Build GRCh38
rs141677716 Chr.17: 30835358-30835358 on Build GRCh38
rs577468040 Chr.17: 30835457-30835457 on Build GRCh38
rs768171111 Chr.17: 30835421-30835421 on Build GRCh38
rs112795423 Chr.17: 30835441-30835441 on Build GRCh38
rs756779040 Chr.17: 30835364-30835364 on Build GRCh38
rs890084111 Chr.17: 30835360-30835360 on Build GRCh38
rs759686535 Chr.17: 30835393-30835393 on Build GRCh38
rs1039880026 Chr.17: 30835268-30835268 on Build GRCh38
rs774562306 Chr.17: 30835267-30835267 on Build GRCh38
rs762445169 Chr.17: 30835259-30835259 on Build GRCh38
rs779103209 Chr.17: 30835334-30835334 on Build GRCh38
rs141483440 Chr.17: 30835322-30835322 on Build GRCh38

View Full Product Details