Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35142290-35142549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDEM2
Entrez Gene ID
55741
Gene Name
ER degradation enhancing alpha-mannosidase like protein 2
Gene Aliases
C20orf31, C20orf49, bA4204.1
UniGene
Hs.720177
Gene Chromosome Location
Chr.20: 35115357-35147358 on Build GRCh38
Transcripts
NM_001145025.1 NM_018217.2 NR_026728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4606636 c.191T>C p.V64A Chr.20: 35142423-35142423 on Build GRCh38
4186569 c.350A>T p.E117V Chr.20: 35142387-35142387 on Build GRCh38
4186570 c.227A>T p.E76V Chr.20: 35142387-35142387 on Build GRCh38
4186571 c.349G>T p.E117* Chr.20: 35142388-35142388 on Build GRCh38
4186572 c.226G>T p.E76* Chr.20: 35142388-35142388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771952256 Chr.20: 35142371-35142371 on Build GRCh38
rs750805682 Chr.20: 35142341-35142341 on Build GRCh38
rs773185360 Chr.20: 35142434-35142435 on Build GRCh38
rs371938796 Chr.20: 35142521-35142521 on Build GRCh38
rs373749622 Chr.20: 35142358-35142358 on Build GRCh38
rs764977629 Chr.20: 35142406-35142406 on Build GRCh38
rs532778574 Chr.20: 35142391-35142391 on Build GRCh38
rs761308105 Chr.20: 35142517-35142517 on Build GRCh38
rs184063388 Chr.20: 35142494-35142494 on Build GRCh38
rs780214317 Chr.20: 35142349-35142349 on Build GRCh38
rs755229780 Chr.20: 35142443-35142443 on Build GRCh38
rs776512001 Chr.20: 35142392-35142392 on Build GRCh38
rs748268998 Chr.20: 35142447-35142447 on Build GRCh38
rs775184458 Chr.20: 35142411-35142411 on Build GRCh38
rs758534825 Chr.20: 35142450-35142450 on Build GRCh38
rs141998071 Chr.20: 35142385-35142385 on Build GRCh38
rs980703414 Chr.20: 35142510-35142510 on Build GRCh38
rs189031450 Chr.20: 35142497-35142497 on Build GRCh38
rs759321315 Chr.20: 35142404-35142404 on Build GRCh38
rs540044847 Chr.20: 35142356-35142356 on Build GRCh38
rs144209865 Chr.20: 35142424-35142424 on Build GRCh38
rs748195066 Chr.20: 35142364-35142364 on Build GRCh38
rs754249409 Chr.20: 35142526-35142526 on Build GRCh38
rs776635644 Chr.20: 35142493-35142493 on Build GRCh38
rs1019039440 Chr.20: 35142499-35142499 on Build GRCh38
rs760773345 Chr.20: 35142487-35142487 on Build GRCh38
rs892524081 Chr.20: 35142377-35142377 on Build GRCh38
rs773717513 Chr.20: 35142511-35142511 on Build GRCh38
rs147639867 Chr.20: 35142381-35142381 on Build GRCh38
rs140217956 Chr.20: 35142446-35142446 on Build GRCh38
rs375577548 Chr.20: 35142360-35142360 on Build GRCh38
rs763671919 Chr.20: 35142500-35142500 on Build GRCh38
rs997048963 Chr.20: 35142481-35142481 on Build GRCh38
rs369596170 Chr.20: 35142335-35142335 on Build GRCh38
rs1056887525 Chr.20: 35142372-35142372 on Build GRCh38
rs978051693 Chr.20: 35142436-35142436 on Build GRCh38
rs917861015 Chr.20: 35142505-35142505 on Build GRCh38
rs770765592 Chr.20: 35142388-35142388 on Build GRCh38
rs777646292 Chr.20: 35142452-35142452 on Build GRCh38
rs891347972 Chr.20: 35142370-35142370 on Build GRCh38
rs369753267 Chr.20: 35142421-35142421 on Build GRCh38
rs370930437 Chr.20: 35142496-35142496 on Build GRCh38
rs763466253 Chr.20: 35142414-35142414 on Build GRCh38
rs766916281 Chr.20: 35142495-35142495 on Build GRCh38
rs151130172 Chr.20: 35142425-35142425 on Build GRCh38
rs150074155 Chr.20: 35142527-35142527 on Build GRCh38
rs947938828 Chr.20: 35142509-35142509 on Build GRCh38
rs920436570 Chr.20: 35142333-35142333 on Build GRCh38
rs756526754 Chr.20: 35142344-35142344 on Build GRCh38
rs762505697 Chr.20: 35142413-35142413 on Build GRCh38
rs376269911 Chr.20: 35142468-35142468 on Build GRCh38
rs556734937 Chr.20: 35142375-35142375 on Build GRCh38
rs375028628 Chr.20: 35142464-35142464 on Build GRCh38
rs375436239 Chr.20: 35142337-35142337 on Build GRCh38
rs756517367 Chr.20: 35142423-35142423 on Build GRCh38

View Full Product Details