Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13209275-13209546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1A
Entrez Gene ID
773
Gene Name
calcium voltage-gated channel subunit alpha1 A
Gene Aliases
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6
UniGene
Hs.501632
Gene Chromosome Location
Chr.19: 13206442-13506460 on Build GRCh38
Transcripts
NM_000068.3 NM_001127221.1 NM_001127222.1 NM_001174080.1 NM_023035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589980 c.6387G>A p.L2129L Chr.19: 13209454-13209454 on Build GRCh38
3937874 c.6455C>G p.S2152C Chr.19: 13209350-13209350 on Build GRCh38
4621057 c.6448C>T p.R2150W Chr.19: 13209390-13209390 on Build GRCh38
6617336 c.6475C>T p.R2159C Chr.19: 13209366-13209366 on Build GRCh38
3937873 c.6488C>G p.S2163C Chr.19: 13209350-13209350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754334160 Chr.19: 13209336-13209336 on Build GRCh38
rs16049 Chr.19: 13209496-13209496 on Build GRCh38
rs370289732 Chr.19: 13209324-13209324 on Build GRCh38
rs965366759 Chr.19: 13209416-13209416 on Build GRCh38
rs867440129 Chr.19: 13209464-13209464 on Build GRCh38
rs994526133 Chr.19: 13209406-13209406 on Build GRCh38
rs759988082 Chr.19: 13209501-13209501 on Build GRCh38
rs1005361810 Chr.19: 13209387-13209387 on Build GRCh38
rs769503871 Chr.19: 13209356-13209356 on Build GRCh38
rs765842344 Chr.19: 13209385-13209385 on Build GRCh38
rs756597852 Chr.19: 13209325-13209325 on Build GRCh38
rs753230279 Chr.19: 13209526-13209526 on Build GRCh38
rs779044548 Chr.19: 13209440-13209440 on Build GRCh38
rs965835901 Chr.19: 13209303-13209303 on Build GRCh38
rs766661019 Chr.19: 13209375-13209375 on Build GRCh38
rs867378853 Chr.19: 13209414-13209414 on Build GRCh38
rs750077868 Chr.19: 13209407-13209407 on Build GRCh38
rs759949928 Chr.19: 13209382-13209382 on Build GRCh38
rs991192417 Chr.19: 13209503-13209503 on Build GRCh38
rs749172043 Chr.19: 13209437-13209437 on Build GRCh38
rs972428713 Chr.19: 13209305-13209305 on Build GRCh38
rs746582303 Chr.19: 13209345-13209345 on Build GRCh38
rs754317278 Chr.19: 13209377-13209377 on Build GRCh38
rs727503832 Chr.19: 13209344-13209344 on Build GRCh38
rs368183370 Chr.19: 13209459-13209459 on Build GRCh38
rs984050286 Chr.19: 13209341-13209341 on Build GRCh38
rs866806995 Chr.19: 13209481-13209481 on Build GRCh38
rs745609731 Chr.19: 13209355-13209355 on Build GRCh38
rs757198773 Chr.19: 13209361-13209363 on Build GRCh38
rs538981913 Chr.19: 13209361-13209361 on Build GRCh38
rs767916527 Chr.19: 13209301-13209301 on Build GRCh38
rs781746273 Chr.19: 13209337-13209337 on Build GRCh38
rs756780624 Chr.19: 13209390-13209390 on Build GRCh38
rs989652217 Chr.19: 13209527-13209527 on Build GRCh38
rs983936305 Chr.19: 13209463-13209463 on Build GRCh38
rs368016666 Chr.19: 13209364-13209364 on Build GRCh38
rs745336852 Chr.19: 13209370-13209370 on Build GRCh38
rs779701046 Chr.19: 13209430-13209430 on Build GRCh38
rs961256392 Chr.19: 13209295-13209295 on Build GRCh38
rs375859889 Chr.19: 13209365-13209365 on Build GRCh38
rs375354077 Chr.19: 13209333-13209333 on Build GRCh38
rs760994682 Chr.19: 13209466-13209466 on Build GRCh38
rs927014998 Chr.19: 13209350-13209350 on Build GRCh38
rs17846928 Chr.19: 13209354-13209354 on Build GRCh38
rs755749925 Chr.19: 13209371-13209371 on Build GRCh38
rs866381013 Chr.19: 13209479-13209479 on Build GRCh38
rs916904971 Chr.19: 13209517-13209517 on Build GRCh38
rs371984902 Chr.19: 13209402-13209402 on Build GRCh38
rs16050 Chr.19: 13209460-13209460 on Build GRCh38
rs576057388 Chr.19: 13209492-13209492 on Build GRCh38
rs916868645 Chr.19: 13209330-13209330 on Build GRCh38
rs751044309 Chr.19: 13209389-13209389 on Build GRCh38
rs1042634748 Chr.19: 13209369-13209369 on Build GRCh38
rs755880731 Chr.19: 13209429-13209429 on Build GRCh38
rs572722130 Chr.19: 13209374-13209374 on Build GRCh38
rs554393704 Chr.19: 13209372-13209372 on Build GRCh38
rs121908235 Chr.19: 13209438-13209438 on Build GRCh38
rs867525404 Chr.19: 13209444-13209444 on Build GRCh38
rs780779363 Chr.19: 13209394-13209394 on Build GRCh38
rs762110794 Chr.19: 13209299-13209299 on Build GRCh38
rs774289573 Chr.19: 13209366-13209366 on Build GRCh38
rs765609971 Chr.19: 13209505-13209505 on Build GRCh38

View Full Product Details