Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48653274-48653525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645932 c.805C>T p.L269F Chr.1: 48653371-48653371 on Build GRCh38
5353148 c.779A>G p.Y260C Chr.1: 48653397-48653397 on Build GRCh38
4961081 c.761C>T p.A254V Chr.1: 48653415-48653415 on Build GRCh38
190390 c.795C>T p.I265I Chr.1: 48653381-48653381 on Build GRCh38
3490568 c.795C>T p.I265I Chr.1: 48653381-48653381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057163140 Chr.1: 48653330-48653330 on Build GRCh38
rs183700256 Chr.1: 48653334-48653334 on Build GRCh38
rs376631765 Chr.1: 48653326-48653326 on Build GRCh38
rs764317600 Chr.1: 48653385-48653385 on Build GRCh38
rs771593014 Chr.1: 48653445-48653445 on Build GRCh38
rs760373509 Chr.1: 48653339-48653339 on Build GRCh38
rs1017613254 Chr.1: 48653351-48653351 on Build GRCh38
rs575132944 Chr.1: 48653453-48653453 on Build GRCh38
rs375321264 Chr.1: 48653448-48653448 on Build GRCh38
rs373660005 Chr.1: 48653442-48653442 on Build GRCh38
rs41289226 Chr.1: 48653455-48653455 on Build GRCh38
rs765165380 Chr.1: 48653368-48653368 on Build GRCh38
rs776478916 Chr.1: 48653359-48653359 on Build GRCh38
rs376443097 Chr.1: 48653404-48653404 on Build GRCh38
rs775468073 Chr.1: 48653494-48653494 on Build GRCh38
rs759328258 Chr.1: 48653471-48653471 on Build GRCh38
rs776280167 Chr.1: 48653462-48653462 on Build GRCh38
rs759257152 Chr.1: 48653362-48653362 on Build GRCh38
rs989801506 Chr.1: 48653473-48653473 on Build GRCh38
rs1029813075 Chr.1: 48653389-48653389 on Build GRCh38
rs369451907 Chr.1: 48653411-48653411 on Build GRCh38
rs372467489 Chr.1: 48653472-48653472 on Build GRCh38
rs200088334 Chr.1: 48653380-48653380 on Build GRCh38
rs371805560 Chr.1: 48653387-48653387 on Build GRCh38
rs747642700 Chr.1: 48653325-48653325 on Build GRCh38
rs371497308 Chr.1: 48653321-48653321 on Build GRCh38
rs766152793 Chr.1: 48653350-48653350 on Build GRCh38
rs34134250 Chr.1: 48653457-48653457 on Build GRCh38
rs544951425 Chr.1: 48653403-48653403 on Build GRCh38
rs757617465 Chr.1: 48653398-48653398 on Build GRCh38
rs752601652 Chr.1: 48653375-48653375 on Build GRCh38
rs780566342 Chr.1: 48653421-48653421 on Build GRCh38

View Full Product Details