Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6623391-6623616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2040771 c.8146G>T p.A2716S Chr.11: 6623530-6623530 on Build GRCh38
2040772 c.8108G>A p.G2703D Chr.11: 6623568-6623568 on Build GRCh38
5360555 c.8193C>A p.I2731I Chr.11: 6623483-6623483 on Build GRCh38
1475793 c.8153A>G p.N2718S Chr.11: 6623523-6623523 on Build GRCh38
7115355 c.8170C>T p.L2724F Chr.11: 6623506-6623506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767817840 Chr.11: 6623496-6623496 on Build GRCh38
rs757858922 Chr.11: 6623439-6623439 on Build GRCh38
rs930350686 Chr.11: 6623543-6623543 on Build GRCh38
rs200795986 Chr.11: 6623413-6623413 on Build GRCh38
rs371484662 Chr.11: 6623555-6623555 on Build GRCh38
rs768301472 Chr.11: 6623452-6623452 on Build GRCh38
rs779449552 Chr.11: 6623553-6623553 on Build GRCh38
rs776456642 Chr.11: 6623453-6623453 on Build GRCh38
rs777507099 Chr.11: 6623579-6623579 on Build GRCh38
rs769785326 Chr.11: 6623479-6623479 on Build GRCh38
rs756532731 Chr.11: 6623522-6623522 on Build GRCh38
rs148148252 Chr.11: 6623490-6623490 on Build GRCh38
rs747937249 Chr.11: 6623472-6623472 on Build GRCh38
rs372127346 Chr.11: 6623435-6623435 on Build GRCh38
rs575189604 Chr.11: 6623504-6623504 on Build GRCh38
rs951256878 Chr.11: 6623415-6623415 on Build GRCh38
rs373867079 Chr.11: 6623445-6623445 on Build GRCh38
rs778420931 Chr.11: 6623429-6623429 on Build GRCh38
rs1044655957 Chr.11: 6623499-6623499 on Build GRCh38
rs150262490 Chr.11: 6623557-6623557 on Build GRCh38
rs766402825 Chr.11: 6623487-6623487 on Build GRCh38
rs746670481 Chr.11: 6623451-6623451 on Build GRCh38
rs754399945 Chr.11: 6623528-6623528 on Build GRCh38
rs558180114 Chr.11: 6623575-6623575 on Build GRCh38
rs1040630141 Chr.11: 6623581-6623581 on Build GRCh38
rs757739092 Chr.11: 6623533-6623533 on Build GRCh38
rs141243126 Chr.11: 6623412-6623412 on Build GRCh38
rs866643933 Chr.11: 6623506-6623506 on Build GRCh38
rs201953922 Chr.11: 6623483-6623483 on Build GRCh38
rs917189635 Chr.11: 6623482-6623482 on Build GRCh38
rs141056372 Chr.11: 6623517-6623517 on Build GRCh38
rs72911012 Chr.11: 6623570-6623570 on Build GRCh38
rs74850790 Chr.11: 6623480-6623480 on Build GRCh38
rs764493359 Chr.11: 6623525-6623525 on Build GRCh38
rs377610020 Chr.11: 6623498-6623498 on Build GRCh38
rs756725661 Chr.11: 6623421-6623421 on Build GRCh38

View Full Product Details