Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71383278-71383545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
UniGene
Hs.106857
Gene Chromosome Location
Chr.16: 71358713-71390438 on Build GRCh38
Transcripts
NM_001740.4 NM_007088.3 NR_027910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6618973 c.431A>G p.N144S Chr.16: 71383398-71383398 on Build GRCh38
4548253 c.442G>A p.D148N Chr.16: 71383409-71383409 on Build GRCh38
6145108 c.414C>A p.D138E Chr.16: 71383381-71383381 on Build GRCh38
3275830 c.433C>T p.R145W Chr.16: 71383400-71383400 on Build GRCh38
1563510 c.438G>A p.P146P Chr.16: 71383405-71383405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374243044 Chr.16: 71383393-71383393 on Build GRCh38
rs769361644 Chr.16: 71383409-71383409 on Build GRCh38
rs779128346 Chr.16: 71383396-71383396 on Build GRCh38
rs143234572 Chr.16: 71383443-71383443 on Build GRCh38
rs567513210 Chr.16: 71383420-71383420 on Build GRCh38
rs769003219 Chr.16: 71383320-71383320 on Build GRCh38
rs748416874 Chr.16: 71383399-71383399 on Build GRCh38
rs776152335 Chr.16: 71383334-71383334 on Build GRCh38
rs773593097 Chr.16: 71383331-71383331 on Build GRCh38
rs781115969 Chr.16: 71383386-71383386 on Build GRCh38
rs746414640 Chr.16: 71383494-71383494 on Build GRCh38
rs756082928 Chr.16: 71383456-71383456 on Build GRCh38
rs1049997903 Chr.16: 71383455-71383455 on Build GRCh38
rs753997772 Chr.16: 71383353-71383354 on Build GRCh38
rs114651335 Chr.16: 71383411-71383411 on Build GRCh38
rs111370553 Chr.16: 71383330-71383330 on Build GRCh38
rs758582606 Chr.16: 71383475-71383475 on Build GRCh38
rs752404823 Chr.16: 71383350-71383350 on Build GRCh38
rs138940173 Chr.16: 71383491-71383491 on Build GRCh38
rs74025974 Chr.16: 71383476-71383476 on Build GRCh38
rs932879862 Chr.16: 71383454-71383454 on Build GRCh38
rs745487693 Chr.16: 71383520-71383520 on Build GRCh38
rs750448570 Chr.16: 71383447-71383447 on Build GRCh38
rs778004370 Chr.16: 71383339-71383339 on Build GRCh38
rs370989254 Chr.16: 71383322-71383322 on Build GRCh38
rs761608945 Chr.16: 71383436-71383436 on Build GRCh38
rs751347567 Chr.16: 71383370-71383370 on Build GRCh38
rs762563993 Chr.16: 71383414-71383414 on Build GRCh38
rs904672519 Chr.16: 71383461-71383461 on Build GRCh38
rs369266349 Chr.16: 71383336-71383336 on Build GRCh38
rs553312346 Chr.16: 71383492-71383492 on Build GRCh38
rs752772941 Chr.16: 71383471-71383471 on Build GRCh38
rs374954837 Chr.16: 71383483-71383483 on Build GRCh38
rs747351423 Chr.16: 71383477-71383477 on Build GRCh38
rs200120146 Chr.16: 71383323-71383323 on Build GRCh38
rs551329852 Chr.16: 71383332-71383332 on Build GRCh38
rs201550865 Chr.16: 71383333-71383333 on Build GRCh38
rs760026354 Chr.16: 71383405-71383405 on Build GRCh38
rs765453701 Chr.16: 71383459-71383459 on Build GRCh38
rs868340522 Chr.16: 71383427-71383427 on Build GRCh38
rs371201286 Chr.16: 71383351-71383351 on Build GRCh38
rs757170928 Chr.16: 71383384-71383384 on Build GRCh38
rs758983810 Chr.16: 71383335-71383335 on Build GRCh38
rs138229238 Chr.16: 71383404-71383404 on Build GRCh38
rs763837619 Chr.16: 71383364-71383364 on Build GRCh38
rs778951263 Chr.16: 71383490-71383491 on Build GRCh38
rs774019948 Chr.16: 71383425-71383425 on Build GRCh38
rs200527047 Chr.16: 71383401-71383401 on Build GRCh38
rs755165733 Chr.16: 71383395-71383395 on Build GRCh38
rs368881886 Chr.16: 71383349-71383349 on Build GRCh38
rs971044789 Chr.16: 71383310-71383310 on Build GRCh38
rs772312901 Chr.16: 71383400-71383400 on Build GRCh38

View Full Product Details