Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10115745-10116000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRK1
Entrez Gene ID
55845
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Gene Aliases
C3orf10, HSPC300, MDS027, hHBrk1
UniGene
Hs.649307
Gene Chromosome Location
Chr.3: 10115649-10127190 on Build GRCh38
Transcripts
NM_018462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3584703 c.117C>T p.F39F Chr.3: 10115818-10115818 on Build GRCh38
4666180 c.72A>G p.I24M Chr.3: 10115773-10115773 on Build GRCh38
4666181 c.72A>G p.I24M Chr.3: 10115773-10115773 on Build GRCh38
445225 c.55C>T p.R19W Chr.3: 10115756-10115756 on Build GRCh38
4648636 c.117C>T p.F39F Chr.3: 10115818-10115818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757747590 Chr.3: 10115863-10115863 on Build GRCh38
rs762409539 Chr.3: 10115940-10115940 on Build GRCh38
rs1044454293 Chr.3: 10115896-10115896 on Build GRCh38
rs994553628 Chr.3: 10115971-10115971 on Build GRCh38
rs928907742 Chr.3: 10115930-10115930 on Build GRCh38
rs957461744 Chr.3: 10115832-10115832 on Build GRCh38
rs917365844 Chr.3: 10115846-10115846 on Build GRCh38
rs752882082 Chr.3: 10115848-10115848 on Build GRCh38
rs28532206 Chr.3: 10115914-10115914 on Build GRCh38
rs781432308 Chr.3: 10115844-10115844 on Build GRCh38
rs28364607 Chr.3: 10115764-10115764 on Build GRCh38
rs552644461 Chr.3: 10115843-10115843 on Build GRCh38
rs897543663 Chr.3: 10115963-10115963 on Build GRCh38
rs372809889 Chr.3: 10115927-10115927 on Build GRCh38
rs915925089 Chr.3: 10115872-10115872 on Build GRCh38
rs777833946 Chr.3: 10115852-10115852 on Build GRCh38
rs750321983 Chr.3: 10115831-10115831 on Build GRCh38
rs766486318 Chr.3: 10115842-10115842 on Build GRCh38
rs939198168 Chr.3: 10115829-10115829 on Build GRCh38
rs923766988 Chr.3: 10115818-10115818 on Build GRCh38
rs148884975 Chr.3: 10115927-10115928 on Build GRCh38
rs1009045387 Chr.3: 10115969-10115969 on Build GRCh38
rs948912481 Chr.3: 10115910-10115910 on Build GRCh38
rs990122778 Chr.3: 10115854-10115854 on Build GRCh38
rs978419249 Chr.3: 10115771-10115771 on Build GRCh38
rs774756890 Chr.3: 10115929-10115929 on Build GRCh38
rs564385275 Chr.3: 10115851-10115851 on Build GRCh38
rs762982724 Chr.3: 10115835-10115835 on Build GRCh38
rs886586487 Chr.3: 10115952-10115952 on Build GRCh38
rs749434405 Chr.3: 10115853-10115853 on Build GRCh38

View Full Product Details