Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1242761-1242988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM132A
Entrez Gene ID
388581
Gene Name
family with sequence similarity 132 member A
Gene Aliases
C1QDC2, C1QTNF12, CTRP12
UniGene
Hs.197613
Gene Chromosome Location
Chr.1: 1242446-1247218 on Build GRCh38
Transcripts
NM_001014980.2 XM_011541435.2 XM_011541436.2 XM_011541437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641442 c.784C>T p.Q262* Chr.1: 1242861-1242861 on Build GRCh38
894517 c.735C>T p.C245C Chr.1: 1242910-1242910 on Build GRCh38
5694413 c.810+2T>A p.? Chr.1: 1242833-1242833 on Build GRCh38
1747662 c.780G>A p.T260T Chr.1: 1242865-1242865 on Build GRCh38
1294791 c.757G>C p.E253Q Chr.1: 1242888-1242888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74511182 Chr.1: 1242864-1242864 on Build GRCh38
rs767386845 Chr.1: 1242962-1242962 on Build GRCh38
rs766564916 Chr.1: 1242945-1242945 on Build GRCh38
rs999021655 Chr.1: 1242800-1242800 on Build GRCh38
rs752550553 Chr.1: 1242826-1242826 on Build GRCh38
rs772407404 Chr.1: 1242910-1242910 on Build GRCh38
rs746855385 Chr.1: 1242818-1242818 on Build GRCh38
rs142136905 Chr.1: 1242835-1242835 on Build GRCh38
rs777616461 Chr.1: 1242815-1242815 on Build GRCh38
rs368046163 Chr.1: 1242799-1242799 on Build GRCh38
rs11260566 Chr.1: 1242865-1242865 on Build GRCh38
rs750669728 Chr.1: 1242863-1242863 on Build GRCh38
rs748763798 Chr.1: 1242896-1242896 on Build GRCh38
rs755775995 Chr.1: 1242782-1242782 on Build GRCh38
rs764049219 Chr.1: 1242951-1242951 on Build GRCh38
rs376266798 Chr.1: 1242792-1242792 on Build GRCh38
rs369515180 Chr.1: 1242901-1242901 on Build GRCh38
rs755990830 Chr.1: 1242837-1242837 on Build GRCh38
rs891290043 Chr.1: 1242926-1242926 on Build GRCh38
rs1023490607 Chr.1: 1242827-1242827 on Build GRCh38
rs774910034 Chr.1: 1242828-1242828 on Build GRCh38
rs757149209 Chr.1: 1242874-1242874 on Build GRCh38
rs182102046 Chr.1: 1242812-1242812 on Build GRCh38
rs760886471 Chr.1: 1242934-1242934 on Build GRCh38
rs369998821 Chr.1: 1242875-1242875 on Build GRCh38
rs762119912 Chr.1: 1242909-1242909 on Build GRCh38
rs757308731 Chr.1: 1242961-1242961 on Build GRCh38
rs148322179 Chr.1: 1242871-1242871 on Build GRCh38
rs779604214 Chr.1: 1242892-1242892 on Build GRCh38
rs1011302055 Chr.1: 1242801-1242801 on Build GRCh38
rs138991752 Chr.1: 1242860-1242860 on Build GRCh38
rs773609972 Chr.1: 1242929-1242929 on Build GRCh38
rs77651834 Chr.1: 1242886-1242886 on Build GRCh38
rs751563244 Chr.1: 1242953-1242953 on Build GRCh38
rs377138690 Chr.1: 1242866-1242866 on Build GRCh38
rs201644366 Chr.1: 1242900-1242900 on Build GRCh38
rs777581156 Chr.1: 1242873-1242873 on Build GRCh38
rs767686793 Chr.1: 1242862-1242862 on Build GRCh38
rs370391953 Chr.1: 1242795-1242795 on Build GRCh38
rs374909487 Chr.1: 1242791-1242791 on Build GRCh38
rs745329919 Chr.1: 1242822-1242822 on Build GRCh38
rs374646388 Chr.1: 1242821-1242821 on Build GRCh38
rs749012617 Chr.1: 1242884-1242884 on Build GRCh38
rs759535031 Chr.1: 1242950-1242950 on Build GRCh38
rs754419886 Chr.1: 1242864-1242864 on Build GRCh38
rs558132433 Chr.1: 1242802-1242802 on Build GRCh38
rs533260195 Chr.1: 1242946-1242946 on Build GRCh38
rs778406520 Chr.1: 1242957-1242957 on Build GRCh38
rs763484176 Chr.1: 1242832-1242832 on Build GRCh38

View Full Product Details