Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 41225067-41225336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
UniGene
Hs.476018
Gene Chromosome Location
Chr.3: 41199451-41240448 on Build GRCh38
Transcripts
NM_001098209.1 NM_001098210.1 NM_001904.3 XM_005264886.2 XM_006712983.1 XM_006712984.1 XM_006712985.1 XM_017005738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6941850 c.102_416del515 p.? Chr.3: 41224614-41225128 on Build GRCh38
6035 c.14_430del417 p.A5_Q143del Chr.3: 41224526-41225142 on Build GRCh38
446497 c.390G>A p.Q130Q Chr.3: 41225102-41225102 on Build GRCh38
6964429 c.367C>T p.Q123* Chr.3: 41225079-41225079 on Build GRCh38
396036 c.419T>C p.I140T Chr.3: 41225131-41225131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779572121 Chr.3: 41225287-41225287 on Build GRCh38
rs771673118 Chr.3: 41225285-41225285 on Build GRCh38
rs571252054 Chr.3: 41225224-41225224 on Build GRCh38
rs767618644 Chr.3: 41225229-41225229 on Build GRCh38
rs749833775 Chr.3: 41225129-41225129 on Build GRCh38
rs768566900 Chr.3: 41225219-41225219 on Build GRCh38
rs768314155 Chr.3: 41225295-41225295 on Build GRCh38
rs892189653 Chr.3: 41225262-41225262 on Build GRCh38
rs752945251 Chr.3: 41225093-41225093 on Build GRCh38
rs892322076 Chr.3: 41225148-41225148 on Build GRCh38
rs759695464 Chr.3: 41225227-41225227 on Build GRCh38
rs764423424 Chr.3: 41225215-41225215 on Build GRCh38
rs201364335 Chr.3: 41225223-41225223 on Build GRCh38
rs202217100 Chr.3: 41225094-41225094 on Build GRCh38
rs775491694 Chr.3: 41225106-41225106 on Build GRCh38
rs537794740 Chr.3: 41225236-41225236 on Build GRCh38
rs760938761 Chr.3: 41225298-41225299 on Build GRCh38
rs60908956 Chr.3: 41225259-41225259 on Build GRCh38
rs3856746 Chr.3: 41225132-41225132 on Build GRCh38
rs778138109 Chr.3: 41225108-41225108 on Build GRCh38
rs753029334 Chr.3: 41225235-41225235 on Build GRCh38
rs771554085 Chr.3: 41225187-41225187 on Build GRCh38
rs778248964 Chr.3: 41225238-41225238 on Build GRCh38
rs903673902 Chr.3: 41225217-41225217 on Build GRCh38
rs369510063 Chr.3: 41225195-41225195 on Build GRCh38
rs1052240748 Chr.3: 41225237-41225237 on Build GRCh38
rs200968230 Chr.3: 41225164-41225164 on Build GRCh38
rs746718978 Chr.3: 41225291-41225291 on Build GRCh38
rs997230964 Chr.3: 41225293-41225293 on Build GRCh38
rs754218841 Chr.3: 41225239-41225239 on Build GRCh38
rs148796283 Chr.3: 41225307-41225307 on Build GRCh38
rs757717193 Chr.3: 41225248-41225248 on Build GRCh38
rs774085540 Chr.3: 41225174-41225174 on Build GRCh38
rs5743392 Chr.3: 41225198-41225198 on Build GRCh38
rs779395096 Chr.3: 41225252-41225252 on Build GRCh38
rs745411917 Chr.3: 41225180-41225180 on Build GRCh38
rs776682401 Chr.3: 41225221-41225221 on Build GRCh38

View Full Product Details