Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 2949322-2949577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
UniGene
Hs.571466
Gene Chromosome Location
Chr.8: 2935353-4995035 on Build GRCh38
Transcripts
NM_033225.5 XM_011534752.2 XM_011534753.2 XM_011534754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3647924 c.9559G>A p.D3187N Chr.8: 2949329-2949329 on Build GRCh38
4519957 c.10366G>A p.G3456R Chr.8: 2949335-2949335 on Build GRCh38
5874503 c.10350T>G p.F3450L Chr.8: 2949351-2949351 on Build GRCh38
3647922 c.9565G>A p.G3189R Chr.8: 2949323-2949323 on Build GRCh38
5733391 c.9566G>T p.G3189V Chr.8: 2949322-2949322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970373709 Chr.8: 2949519-2949519 on Build GRCh38
rs772236278 Chr.8: 2949386-2949386 on Build GRCh38
rs766284756 Chr.8: 2949396-2949400 on Build GRCh38
rs994370101 Chr.8: 2949451-2949451 on Build GRCh38
rs5741961 Chr.8: 2949398-2949399 on Build GRCh38
rs199630799 Chr.8: 2949416-2949416 on Build GRCh38
rs559219001 Chr.8: 2949428-2949428 on Build GRCh38
rs1008282390 Chr.8: 2949440-2949440 on Build GRCh38
rs748212272 Chr.8: 2949379-2949379 on Build GRCh38
rs145997196 Chr.8: 2949411-2949412 on Build GRCh38
rs752253287 Chr.8: 2949426-2949426 on Build GRCh38
rs368841292 Chr.8: 2949402-2949403 on Build GRCh38
rs750088444 Chr.8: 2949407-2949408 on Build GRCh38
rs764758136 Chr.8: 2949425-2949425 on Build GRCh38
rs1105081 Chr.8: 2949530-2949530 on Build GRCh38
rs775636267 Chr.8: 2949544-2949545 on Build GRCh38
rs762511500 Chr.8: 2949427-2949427 on Build GRCh38
rs57461657 Chr.8: 2949415-2949416 on Build GRCh38
rs372864008 Chr.8: 2949370-2949370 on Build GRCh38
rs762953817 Chr.8: 2949373-2949373 on Build GRCh38
rs201743560 Chr.8: 2949548-2949548 on Build GRCh38
rs962075493 Chr.8: 2949499-2949499 on Build GRCh38
rs754463929 Chr.8: 2949408-2949409 on Build GRCh38
rs188927899 Chr.8: 2949359-2949359 on Build GRCh38
rs73657526 Chr.8: 2949398-2949398 on Build GRCh38
rs989302227 Chr.8: 2949547-2949547 on Build GRCh38
rs201340269 Chr.8: 2949424-2949424 on Build GRCh38
rs376344419 Chr.8: 2949547-2949550 on Build GRCh38
rs539190756 Chr.8: 2949395-2949395 on Build GRCh38
rs796862142 Chr.8: 2949476-2949476 on Build GRCh38
rs72288499 Chr.8: 2949545-2949548 on Build GRCh38
rs777251815 Chr.8: 2949399-2949400 on Build GRCh38
rs144666650 Chr.8: 2949501-2949501 on Build GRCh38
rs756445765 Chr.8: 2949432-2949432 on Build GRCh38
rs768925991 Chr.8: 2949416-2949417 on Build GRCh38
rs751155197 Chr.8: 2949429-2949429 on Build GRCh38
rs140807526 Chr.8: 2949443-2949443 on Build GRCh38
rs1008943083 Chr.8: 2949480-2949480 on Build GRCh38
rs749751514 Chr.8: 2949364-2949364 on Build GRCh38
rs897314024 Chr.8: 2949447-2949447 on Build GRCh38
rs773472772 Chr.8: 2949392-2949392 on Build GRCh38
rs779091673 Chr.8: 2949371-2949371 on Build GRCh38
rs778590309 Chr.8: 2949399-2949403 on Build GRCh38
rs746191318 Chr.8: 2949412-2949413 on Build GRCh38
rs1003822096 Chr.8: 2949522-2949522 on Build GRCh38
rs200448663 Chr.8: 2949417-2949417 on Build GRCh38
rs541490978 Chr.8: 2949477-2949477 on Build GRCh38
rs760689584 Chr.8: 2949395-2949396 on Build GRCh38
rs1014592179 Chr.8: 2949496-2949496 on Build GRCh38
rs200675652 Chr.8: 2949423-2949423 on Build GRCh38
rs267601885 Chr.8: 2949367-2949367 on Build GRCh38
rs543462035 Chr.8: 2949523-2949523 on Build GRCh38
rs969177735 Chr.8: 2949532-2949532 on Build GRCh38
rs1023216412 Chr.8: 2949502-2949502 on Build GRCh38

View Full Product Details