Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130565462-130565703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A6
Entrez Gene ID
131873
Gene Name
collagen type VI alpha 6
Gene Aliases
-
UniGene
Hs.591282
Gene Chromosome Location
Chr.3: 130516709-130678135 on Build GRCh38
Transcripts
NM_001102608.1 XM_005247121.4 XM_011512423.2 XM_011512424.2 XM_011512425.2 XM_011512426.2 XM_011512428.2 XM_011512431.2 XM_011512432.2 XM_017005711.1 XM_017005712.1 XM_017005713.1 XM_017005714.1 XM_017005715.1 XM_017005716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522665 c.1173G>A p.K391K Chr.3: 130565505-130565505 on Build GRCh38
1202019 c.1271C>T p.T424M Chr.3: 130565603-130565603 on Build GRCh38
6987882 c.1170G>T p.L390L Chr.3: 130565502-130565502 on Build GRCh38
6529024 c.1266T>C p.T422T Chr.3: 130565598-130565598 on Build GRCh38
4931157 c.1153C>A p.Q385K Chr.3: 130565485-130565485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747936555 Chr.3: 130565540-130565540 on Build GRCh38
rs774243865 Chr.3: 130565537-130565537 on Build GRCh38
rs765967672 Chr.3: 130565562-130565562 on Build GRCh38
rs763551831 Chr.3: 130565579-130565579 on Build GRCh38
rs756277929 Chr.3: 130565621-130565621 on Build GRCh38
rs1018348763 Chr.3: 130565652-130565654 on Build GRCh38
rs867627022 Chr.3: 130565602-130565602 on Build GRCh38
rs775997763 Chr.3: 130565568-130565568 on Build GRCh38
rs751941965 Chr.3: 130565601-130565601 on Build GRCh38
rs372038797 Chr.3: 130565512-130565512 on Build GRCh38
rs199714543 Chr.3: 130565555-130565555 on Build GRCh38
rs957347009 Chr.3: 130565531-130565531 on Build GRCh38
rs1006954867 Chr.3: 130565651-130565652 on Build GRCh38
rs200519575 Chr.3: 130565615-130565615 on Build GRCh38
rs200576820 Chr.3: 130565603-130565603 on Build GRCh38
rs760270545 Chr.3: 130565561-130565561 on Build GRCh38
rs528723799 Chr.3: 130565604-130565604 on Build GRCh38
rs955244518 Chr.3: 130565598-130565598 on Build GRCh38
rs770189741 Chr.3: 130565554-130565554 on Build GRCh38
rs749299091 Chr.3: 130565664-130565664 on Build GRCh38
rs941188150 Chr.3: 130565524-130565524 on Build GRCh38
rs764484060 Chr.3: 130565585-130565585 on Build GRCh38
rs768475459 Chr.3: 130565534-130565534 on Build GRCh38
rs891126372 Chr.3: 130565637-130565641 on Build GRCh38
rs369911831 Chr.3: 130565595-130565595 on Build GRCh38
rs377462836 Chr.3: 130565660-130565660 on Build GRCh38
rs202062422 Chr.3: 130565511-130565511 on Build GRCh38
rs547523404 Chr.3: 130565525-130565525 on Build GRCh38
rs942567132 Chr.3: 130565656-130565656 on Build GRCh38
rs745976897 Chr.3: 130565504-130565504 on Build GRCh38
rs777130542 Chr.3: 130565600-130565600 on Build GRCh38
rs973841110 Chr.3: 130565680-130565680 on Build GRCh38

View Full Product Details