Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46734786-46735055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7B2
Entrez Gene ID
170712
Gene Name
cytochrome c oxidase subunit 7B2
Gene Aliases
-
UniGene
Hs.479656
Gene Chromosome Location
Chr.4: 46734827-46909266 on Build GRCh38
Transcripts
NM_130902.2 XM_005248056.4 XM_011513630.2 XM_011513631.2 XM_011513632.2 XM_011513633.2 XM_011513634.2 XM_011513635.2 XM_011513636.1 XM_011513637.2 XM_011513638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1055674 c.199C>A p.L67I Chr.4: 46734994-46734994 on Build GRCh38
5401239 c.166T>A p.F56I Chr.4: 46735027-46735027 on Build GRCh38
5716717 c.194G>A p.W65* Chr.4: 46734999-46734999 on Build GRCh38
1727945 c.224C>A p.P75Q Chr.4: 46734969-46734969 on Build GRCh38
7096049 c.147C>G p.F49L Chr.4: 46735046-46735046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369665627 Chr.4: 46734995-46734995 on Build GRCh38
rs562719730 Chr.4: 46734853-46734853 on Build GRCh38
rs566933835 Chr.4: 46734913-46734913 on Build GRCh38
rs1002773641 Chr.4: 46734985-46734985 on Build GRCh38
rs141223274 Chr.4: 46734860-46734860 on Build GRCh38
rs753443824 Chr.4: 46734984-46734984 on Build GRCh38
rs750184358 Chr.4: 46734970-46734970 on Build GRCh38
rs781693484 Chr.4: 46734986-46734987 on Build GRCh38
rs371845383 Chr.4: 46734918-46734918 on Build GRCh38
rs552803258 Chr.4: 46734958-46734958 on Build GRCh38
rs759649183 Chr.4: 46734948-46734948 on Build GRCh38
rs766001089 Chr.4: 46734981-46734981 on Build GRCh38
rs868077904 Chr.4: 46734897-46734897 on Build GRCh38
rs1039658163 Chr.4: 46734852-46734852 on Build GRCh38
rs933857579 Chr.4: 46734855-46734855 on Build GRCh38
rs201135842 Chr.4: 46735024-46735024 on Build GRCh38
rs571204159 Chr.4: 46734963-46734963 on Build GRCh38
rs770332163 Chr.4: 46734886-46734886 on Build GRCh38
rs548408963 Chr.4: 46734861-46734861 on Build GRCh38
rs375968257 Chr.4: 46734941-46734941 on Build GRCh38
rs747374356 Chr.4: 46735022-46735022 on Build GRCh38
rs778464197 Chr.4: 46735011-46735011 on Build GRCh38
rs767255171 Chr.4: 46734969-46734969 on Build GRCh38
rs905302386 Chr.4: 46734950-46734950 on Build GRCh38
rs550141419 Chr.4: 46734870-46734870 on Build GRCh38
rs912353780 Chr.4: 46734843-46734843 on Build GRCh38
rs775138297 Chr.4: 46734965-46734965 on Build GRCh38
rs1039924660 Chr.4: 46734849-46734849 on Build GRCh38
rs144110097 Chr.4: 46734945-46734945 on Build GRCh38
rs11736008 Chr.4: 46734836-46734836 on Build GRCh38
rs538139494 Chr.4: 46734972-46734972 on Build GRCh38
rs886747256 Chr.4: 46734899-46734899 on Build GRCh38
rs754501633 Chr.4: 46734914-46734914 on Build GRCh38
rs758809331 Chr.4: 46734998-46734998 on Build GRCh38
rs778315894 Chr.4: 46734992-46734992 on Build GRCh38
rs113832521 Chr.4: 46734947-46734947 on Build GRCh38
rs146477410 Chr.4: 46734989-46734989 on Build GRCh38
rs372856051 Chr.4: 46734920-46734920 on Build GRCh38
rs769780584 Chr.4: 46734956-46734956 on Build GRCh38
rs140822119 Chr.4: 46735023-46735023 on Build GRCh38
rs746084227 Chr.4: 46735027-46735027 on Build GRCh38
rs772441556 Chr.4: 46734932-46734932 on Build GRCh38
rs769880381 Chr.4: 46735029-46735029 on Build GRCh38
rs771248119 Chr.4: 46734942-46734942 on Build GRCh38
rs755738219 Chr.4: 46734907-46734907 on Build GRCh38
rs550989099 Chr.4: 46734844-46734844 on Build GRCh38
rs200958105 Chr.4: 46734983-46734983 on Build GRCh38

View Full Product Details