Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 4979592-4979825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHLHE40
Entrez Gene ID
8553
Gene Name
basic helix-loop-helix family member e40
Gene Aliases
BHLHB2, DEC1, HLHB2, SHARP-2, STRA13, Stra14
UniGene
Hs.744856
Gene Chromosome Location
Chr.3: 4979412-4985181 on Build GRCh38
Transcripts
NM_003670.2 NR_037903.3 NR_125915.1 NR_125916.1

Gene Symbol
BHLHE40-AS1
Entrez Gene ID
100507582
Gene Name
BHLHE40 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434505
Gene Chromosome Location
Chr.3: 4896809-4979961 on Build GRCh38
Transcripts
NM_003670.2 NR_037903.3 NR_125915.1 NR_125916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6332826 c.33C>A p.P11P Chr.3: 4979751-4979751 on Build GRCh38
3824206 c.9G>A p.R3R Chr.3: 4979727-4979727 on Build GRCh38
6273647 c.80+5G>T p.? Chr.3: 4979803-4979803 on Build GRCh38
5086452 c.28delC p.A12fs*32 Chr.3: 4979746-4979746 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376351623 Chr.3: 4979711-4979711 on Build GRCh38
rs2271566 Chr.3: 4979718-4979718 on Build GRCh38
rs768885272 Chr.3: 4979702-4979702 on Build GRCh38
rs1005531700 Chr.3: 4979745-4979745 on Build GRCh38
rs999465173 Chr.3: 4979749-4979749 on Build GRCh38
rs1047985243 Chr.3: 4979675-4979675 on Build GRCh38
rs1001794233 Chr.3: 4979720-4979720 on Build GRCh38
rs776510674 Chr.3: 4979742-4979742 on Build GRCh38
rs182209585 Chr.3: 4979696-4979696 on Build GRCh38
rs766430315 Chr.3: 4979804-4979804 on Build GRCh38
rs765080812 Chr.3: 4979784-4979784 on Build GRCh38
rs868723963 Chr.3: 4979746-4979746 on Build GRCh38
rs750412210 Chr.3: 4979787-4979787 on Build GRCh38
rs758941639 Chr.3: 4979713-4979713 on Build GRCh38
rs147293605 Chr.3: 4979714-4979714 on Build GRCh38
rs781492332 Chr.3: 4979710-4979710 on Build GRCh38
rs760364567 Chr.3: 4979725-4979725 on Build GRCh38
rs1046234897 Chr.3: 4979726-4979726 on Build GRCh38
rs763843136 Chr.3: 4979732-4979732 on Build GRCh38
rs565467036 Chr.3: 4979752-4979752 on Build GRCh38
rs758383061 Chr.3: 4979794-4979794 on Build GRCh38
rs906318235 Chr.3: 4979730-4979730 on Build GRCh38
rs1030845006 Chr.3: 4979772-4979772 on Build GRCh38
rs771476449 Chr.3: 4979774-4979774 on Build GRCh38
rs551434839 Chr.3: 4979689-4979689 on Build GRCh38
rs761756861 Chr.3: 4979750-4979750 on Build GRCh38
rs111860035 Chr.3: 4979640-4979640 on Build GRCh38
rs923401766 Chr.3: 4979633-4979633 on Build GRCh38
rs574094733 Chr.3: 4979677-4979677 on Build GRCh38

View Full Product Details