Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158127430-158127665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM198B
Entrez Gene ID
51313
Gene Name
family with sequence similarity 198 member B
Gene Aliases
AD021, AD036, C4orf18
UniGene
Hs.567498
Gene Chromosome Location
Chr.4: 158124580-158173050 on Build GRCh38
Transcripts
NM_001031700.2 NM_001128424.1 NM_016613.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4569074 c.1500T>C p.L500L Chr.4: 158127491-158127491 on Build GRCh38
5671335 c.1458G>T p.V486V Chr.4: 158127533-158127533 on Build GRCh38
1428271 c.1531C>A p.L511I Chr.4: 158127460-158127460 on Build GRCh38
5709132 c.1420C>T p.R474W Chr.4: 158127571-158127571 on Build GRCh38
1428272 c.1507C>A p.L503I Chr.4: 158127460-158127460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761732367 Chr.4: 158127514-158127514 on Build GRCh38
rs1002228467 Chr.4: 158127531-158127531 on Build GRCh38
rs143892979 Chr.4: 158127540-158127540 on Build GRCh38
rs564358793 Chr.4: 158127519-158127519 on Build GRCh38
rs149461557 Chr.4: 158127530-158127530 on Build GRCh38
rs376536173 Chr.4: 158127509-158127509 on Build GRCh38
rs143315826 Chr.4: 158127588-158127588 on Build GRCh38
rs748042605 Chr.4: 158127624-158127624 on Build GRCh38
rs774545799 Chr.4: 158127501-158127501 on Build GRCh38
rs977403422 Chr.4: 158127599-158127599 on Build GRCh38
rs960899360 Chr.4: 158127554-158127554 on Build GRCh38
rs773013456 Chr.4: 158127520-158127520 on Build GRCh38
rs759040738 Chr.4: 158127534-158127534 on Build GRCh38
rs998460273 Chr.4: 158127494-158127494 on Build GRCh38
rs777998205 Chr.4: 158127484-158127484 on Build GRCh38
rs756417549 Chr.4: 158127481-158127481 on Build GRCh38
rs533432929 Chr.4: 158127527-158127527 on Build GRCh38
rs370072337 Chr.4: 158127528-158127528 on Build GRCh38
rs772005578 Chr.4: 158127619-158127619 on Build GRCh38
rs749442476 Chr.4: 158127485-158127485 on Build GRCh38
rs757541102 Chr.4: 158127592-158127592 on Build GRCh38
rs376386222 Chr.4: 158127570-158127570 on Build GRCh38
rs139005471 Chr.4: 158127452-158127452 on Build GRCh38
rs771009632 Chr.4: 158127488-158127488 on Build GRCh38
rs776410355 Chr.4: 158127513-158127513 on Build GRCh38
rs992775697 Chr.4: 158127571-158127571 on Build GRCh38
rs753158894 Chr.4: 158127544-158127544 on Build GRCh38
rs773211304 Chr.4: 158127630-158127630 on Build GRCh38
rs749698008 Chr.4: 158127623-158127623 on Build GRCh38
rs373824929 Chr.4: 158127568-158127568 on Build GRCh38
rs745929444 Chr.4: 158127508-158127508 on Build GRCh38
rs547547781 Chr.4: 158127480-158127480 on Build GRCh38
rs751022476 Chr.4: 158127471-158127471 on Build GRCh38
rs781523565 Chr.4: 158127479-158127479 on Build GRCh38
rs769843062 Chr.4: 158127629-158127629 on Build GRCh38
rs547887790 Chr.4: 158127600-158127600 on Build GRCh38
rs773233579 Chr.4: 158127549-158127550 on Build GRCh38
rs768677634 Chr.4: 158127511-158127511 on Build GRCh38
rs763039055 Chr.4: 158127571-158127572 on Build GRCh38
rs755579740 Chr.4: 158127474-158127474 on Build GRCh38
rs756472594 Chr.4: 158127564-158127564 on Build GRCh38

View Full Product Details