Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129257439-129257712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG1
Entrez Gene ID
22820
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
UniGene
Hs.518250
Gene Chromosome Location
Chr.3: 129249575-129277773 on Build GRCh38
Transcripts
NM_016128.3 XM_011512549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4985732 c.676C>A p.P226T Chr.3: 129257566-129257566 on Build GRCh38
3587183 c.677C>T p.P226L Chr.3: 129257567-129257567 on Build GRCh38
6634066 c.610C>T p.R204C Chr.3: 129257500-129257500 on Build GRCh38
149466 c.663T>C p.H221H Chr.3: 129257553-129257553 on Build GRCh38
7248885 c.718C>G p.L240V Chr.3: 129257608-129257608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781312450 Chr.3: 129257512-129257512 on Build GRCh38
rs748999038 Chr.3: 129257530-129257530 on Build GRCh38
rs373108909 Chr.3: 129257610-129257610 on Build GRCh38
rs370430335 Chr.3: 129257691-129257691 on Build GRCh38
rs753043191 Chr.3: 129257487-129257487 on Build GRCh38
rs200402729 Chr.3: 129257692-129257692 on Build GRCh38
rs756091943 Chr.3: 129257676-129257676 on Build GRCh38
rs761562902 Chr.3: 129257548-129257548 on Build GRCh38
rs907902971 Chr.3: 129257552-129257552 on Build GRCh38
rs935117061 Chr.3: 129257633-129257633 on Build GRCh38
rs747788979 Chr.3: 129257602-129257602 on Build GRCh38
rs746800767 Chr.3: 129257522-129257522 on Build GRCh38
rs760088546 Chr.3: 129257559-129257559 on Build GRCh38
rs763167360 Chr.3: 129257566-129257566 on Build GRCh38
rs773819629 Chr.3: 129257538-129257538 on Build GRCh38
rs959828023 Chr.3: 129257654-129257654 on Build GRCh38
rs974286308 Chr.3: 129257467-129257467 on Build GRCh38
rs765620316 Chr.3: 129257564-129257564 on Build GRCh38
rs916584489 Chr.3: 129257466-129257466 on Build GRCh38
rs753715514 Chr.3: 129257690-129257690 on Build GRCh38
rs200319190 Chr.3: 129257632-129257632 on Build GRCh38
rs764520625 Chr.3: 129257579-129257579 on Build GRCh38
rs370219257 Chr.3: 129257583-129257583 on Build GRCh38
rs779809650 Chr.3: 129257590-129257590 on Build GRCh38
rs768520281 Chr.3: 129257532-129257532 on Build GRCh38
rs758821197 Chr.3: 129257494-129257494 on Build GRCh38
rs756221218 Chr.3: 129257589-129257589 on Build GRCh38
rs200226743 Chr.3: 129257588-129257588 on Build GRCh38
rs113788895 Chr.3: 129257648-129257648 on Build GRCh38
rs888795395 Chr.3: 129257561-129257561 on Build GRCh38
rs768271150 Chr.3: 129257593-129257593 on Build GRCh38
rs778835626 Chr.3: 129257598-129257598 on Build GRCh38
rs763580442 Chr.3: 129257642-129257642 on Build GRCh38
rs545183134 Chr.3: 129257582-129257582 on Build GRCh38
rs775328874 Chr.3: 129257521-129257521 on Build GRCh38
rs762136146 Chr.3: 129257501-129257501 on Build GRCh38
rs4927910 Chr.3: 129257553-129257553 on Build GRCh38
rs747266916 Chr.3: 129257500-129257500 on Build GRCh38
rs767690538 Chr.3: 129257662-129257662 on Build GRCh38
rs62266876 Chr.3: 129257608-129257608 on Build GRCh38
rs751722653 Chr.3: 129257581-129257581 on Build GRCh38
rs765674351 Chr.3: 129257478-129257478 on Build GRCh38
rs531273625 Chr.3: 129257464-129257465 on Build GRCh38
rs764991686 Chr.3: 129257513-129257513 on Build GRCh38
rs372947359 Chr.3: 129257619-129257619 on Build GRCh38
rs780188286 Chr.3: 129257688-129257688 on Build GRCh38
rs767131343 Chr.3: 129257549-129257549 on Build GRCh38
rs1052122356 Chr.3: 129257640-129257640 on Build GRCh38
rs1035528586 Chr.3: 129257649-129257649 on Build GRCh38
rs202155559 Chr.3: 129257637-129257637 on Build GRCh38
rs760213055 Chr.3: 129257612-129257612 on Build GRCh38
rs201721296 Chr.3: 129257520-129257520 on Build GRCh38
rs377347730 Chr.3: 129257667-129257667 on Build GRCh38
rs749264117 Chr.3: 129257591-129257591 on Build GRCh38
rs925413629 Chr.3: 129257497-129257497 on Build GRCh38
rs1043833633 Chr.3: 129257623-129257623 on Build GRCh38
rs751964881 Chr.3: 129257658-129257658 on Build GRCh38
rs755610468 Chr.3: 129257475-129257475 on Build GRCh38
rs761996406 Chr.3: 129257659-129257659 on Build GRCh38

View Full Product Details