Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36253662-36253887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC169
Entrez Gene ID
728591
Gene Name
coiled-coil domain containing 169
Gene Aliases
C13orf38
UniGene
Hs.656745
Gene Chromosome Location
Chr.13: 36227042-36297855 on Build GRCh38
Transcripts
NM_001144981.2 NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001144985.2 NM_001144986.2 NM_001198908.1 NM_001198910.1

Gene Symbol
CCDC169-SOHLH2
Entrez Gene ID
100526761
Gene Name
CCDC169-SOHLH2 readthrough
Gene Aliases
C13orf38-SOHLH2
UniGene
Hs.124519
Gene Chromosome Location
Chr.13: 36168208-36297855 on Build GRCh38
Transcripts
NM_001144981.2 NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001144985.2 NM_001144986.2 NM_001198908.1 NM_001198910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622820 c.444T>C p.R148R Chr.13: 36253827-36253827 on Build GRCh38
5970979 c.140G>T p.R47L Chr.13: 36253831-36253831 on Build GRCh38
4622821 c.144T>C p.R48R Chr.13: 36253827-36253827 on Build GRCh38
5970978 c.440G>T p.R147L Chr.13: 36253831-36253831 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs907828882 Chr.13: 36253760-36253760 on Build GRCh38
rs1001225111 Chr.13: 36253742-36253742 on Build GRCh38
rs1022615259 Chr.13: 36253843-36253843 on Build GRCh38
rs940960264 Chr.13: 36253763-36253763 on Build GRCh38
rs969683806 Chr.13: 36253723-36253723 on Build GRCh38
rs368819109 Chr.13: 36253743-36253743 on Build GRCh38
rs980179870 Chr.13: 36253721-36253721 on Build GRCh38
rs79770535 Chr.13: 36253832-36253832 on Build GRCh38
rs9546886 Chr.13: 36253696-36253696 on Build GRCh38
rs536585049 Chr.13: 36253790-36253791 on Build GRCh38
rs9602561 Chr.13: 36253820-36253820 on Build GRCh38
rs764565358 Chr.13: 36253784-36253784 on Build GRCh38
rs969169819 Chr.13: 36253703-36253703 on Build GRCh38
rs9546887 Chr.13: 36253759-36253759 on Build GRCh38
rs1034383028 Chr.13: 36253737-36253737 on Build GRCh38
rs553550510 Chr.13: 36253829-36253829 on Build GRCh38
rs974034883 Chr.13: 36253776-36253776 on Build GRCh38
rs905063779 Chr.13: 36253850-36253850 on Build GRCh38
rs748021831 Chr.13: 36253860-36253860 on Build GRCh38
rs749890264 Chr.13: 36253804-36253804 on Build GRCh38
rs539132048 Chr.13: 36253837-36253837 on Build GRCh38
rs568355610 Chr.13: 36253796-36253796 on Build GRCh38
rs749538132 Chr.13: 36253829-36253829 on Build GRCh38
rs890203390 Chr.13: 36253824-36253824 on Build GRCh38
rs868484213 Chr.13: 36253831-36253831 on Build GRCh38
rs762444389 Chr.13: 36253795-36253795 on Build GRCh38
rs372590316 Chr.13: 36253793-36253793 on Build GRCh38

View Full Product Details