Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 93828911-93829146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH1
Entrez Gene ID
474
Gene Name
atonal bHLH transcription factor 1
Gene Aliases
ATH1, HATH1, MATH-1, bHLHa14
UniGene
Hs.532680
Gene Chromosome Location
Chr.4: 93828927-93829991 on Build GRCh38
Transcripts
NM_005172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3776078 c.173G>T p.S58I Chr.4: 93829099-93829099 on Build GRCh38
6344864 c.31G>T p.A11S Chr.4: 93828957-93828957 on Build GRCh38
4758947 c.182C>A p.P61Q Chr.4: 93829108-93829108 on Build GRCh38
127383 c.165C>G p.L55L Chr.4: 93829091-93829091 on Build GRCh38
5401837 c.216C>T p.I72I Chr.4: 93829142-93829142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952424149 Chr.4: 93829019-93829019 on Build GRCh38
rs764742976 Chr.4: 93828988-93828988 on Build GRCh38
rs772119264 Chr.4: 93829030-93829030 on Build GRCh38
rs759383481 Chr.4: 93828983-93828983 on Build GRCh38
rs145839632 Chr.4: 93829059-93829059 on Build GRCh38
rs768194059 Chr.4: 93829016-93829017 on Build GRCh38
rs775248036 Chr.4: 93829067-93829067 on Build GRCh38
rs878909771 Chr.4: 93829021-93829021 on Build GRCh38
rs748409858 Chr.4: 93829023-93829023 on Build GRCh38
rs553793713 Chr.4: 93829018-93829018 on Build GRCh38
rs747125484 Chr.4: 93829042-93829042 on Build GRCh38
rs377287614 Chr.4: 93828995-93828995 on Build GRCh38
rs749547808 Chr.4: 93829013-93829013 on Build GRCh38
rs201089265 Chr.4: 93829009-93829009 on Build GRCh38
rs779299219 Chr.4: 93829089-93829089 on Build GRCh38
rs868023894 Chr.4: 93829110-93829110 on Build GRCh38
rs770709819 Chr.4: 93829044-93829044 on Build GRCh38
rs369720969 Chr.4: 93828936-93828936 on Build GRCh38
rs573835433 Chr.4: 93829068-93829068 on Build GRCh38
rs779104212 Chr.4: 93829022-93829022 on Build GRCh38
rs754425733 Chr.4: 93829080-93829080 on Build GRCh38
rs773229402 Chr.4: 93829036-93829036 on Build GRCh38
rs759242809 Chr.4: 93829060-93829060 on Build GRCh38
rs374062652 Chr.4: 93829112-93829112 on Build GRCh38
rs752452091 Chr.4: 93828989-93828989 on Build GRCh38
rs756527546 Chr.4: 93829006-93829006 on Build GRCh38
rs750940636 Chr.4: 93829003-93829003 on Build GRCh38
rs138388359 Chr.4: 93829076-93829076 on Build GRCh38
rs896790063 Chr.4: 93828950-93828950 on Build GRCh38
rs746761979 Chr.4: 93829065-93829065 on Build GRCh38
rs896213120 Chr.4: 93829014-93829014 on Build GRCh38
rs760657587 Chr.4: 93828933-93828933 on Build GRCh38
rs113356228 Chr.4: 93829100-93829100 on Build GRCh38
rs755267235 Chr.4: 93829083-93829083 on Build GRCh38
rs375159413 Chr.4: 93828999-93828999 on Build GRCh38
rs746909201 Chr.4: 93829017-93829019 on Build GRCh38
rs562102963 Chr.4: 93829079-93829079 on Build GRCh38
rs777576567 Chr.4: 93829061-93829061 on Build GRCh38
rs776252021 Chr.4: 93828980-93828980 on Build GRCh38
rs377166186 Chr.4: 93828953-93828953 on Build GRCh38
rs373441827 Chr.4: 93828968-93828968 on Build GRCh38

View Full Product Details